PloS one
Cell wall-anchored nuclease of Streptococcus sanguinis contributes to escape from neutrophil extracellular trap-mediated bacteriocidal activity.
PMID 25084357
Abstract
Streptococcus sanguinis, a member of the commensal mitis group of streptococci, is a primary colonizer of the tooth surface, and has been implicated in infectious complications including bacteremia and infective endocarditis. During disease progression, S. sanguinis may utilize various cell surface molecules to evade the host immune system to survive in blood. In the present study, we discovered a novel cell surface nuclease with a cell-wall anchor domain, termed SWAN (streptococcal wall-anchored nuclease), and investigated its contribution to bacterial resistance against the bacteriocidal activity of neutrophil extracellular traps (NETs). Recombinant SWAN protein (rSWAN) digested multiple forms of DNA including NET DNA and human RNA, which required both Mg(2+) and Ca(2+) for optimum activity. Furthermore, DNase activity of S. sanguinis was detected around growing colonies on agar plates containing DNA. In-frame deletion of the swan gene mostly reduced that activity. These findings indicated that SWAN is a major nuclease displayed on the surface, which was further confirmed by immuno-detection of SWAN in the cell wall fraction. The sensitivity of S. sanguinis to NET killing was reduced by swan gene deletion. Moreover, heterologous expression of the swan gene rendered a Lactococcus lactis strain more resistant to NET killing. Our results suggest that the SWAN nuclease on the bacterial surface contributes to survival in the potential situation of S. sanguinis encountering NETs during the course of disease progression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59349
Ampicillin, analytical standard
C16H19N3O4S
1033000
Ampicillin, United States Pharmacopeia (USP) Reference Standard
C16H19N3O4S
A1000000
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
C16H19N3O4S
C0378
Chloramphenicol, ≥98% (HPLC)
C11H12Cl2N2O5
C7795
Chloramphenicol, γ-irradiated
C11H12Cl2N2O5
C1919
Chloramphenicol, BioReagent, suitable for plant cell culture
C11H12Cl2N2O5
C0857
Chloramphenicol, meets USP testing specifications
C11H12Cl2N2O5
C1200000
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
C11H12Cl2N2O5
31667
Chloramphenicol, VETRANAL, analytical standard
C11H12Cl2N2O5
23276
Chloramphenicol, tested according to Ph.Eur.
C11H12Cl2N2O5
C1863
Chloramphenicol, Biotechnology Performance Certified, suitable for plant cell culture
C11H12Cl2N2O5
C8273
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
C30H37NO6
C2618
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO, 0.2 μm filtered
C30H37NO6
E6376
Erythromycin, potency: ≥850 μg per mg
C37H67NO13
E5389
Erythromycin, BioReagent, suitable for cell culture
C37H67NO13
45674
Erythromycin, tested according to Ph.Eur.
C37H67NO13
PHR1039
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
C37H67NO13
E1300000
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
C37H67NO13
1242000
Erythromycin, United States Pharmacopeia (USP) Reference Standard
C37H67NO13
E0774
Erythromycin, meets USP testing specifications
C37H67NO13
E4514
Erythromycin, plant cell culture tested, ~98%
C37H67NO13
45703
Erythromycin standard solution, 1 mg/mL in H2O
C37H67NO13