EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
ELF-MF attenuates quercetin-induced apoptosis in K562 cells through modulating the expression of Bcl-2 family proteins.
PMID 25084985
Abstract
This study investigated the effects of sinusoidal ELF-MF (1xa0mT; 50xa0Hz) on the apoptosis induced by four different compounds, namely vinblastine, etoposide, quercetin, and resveratrol, in human K562 chronic myeloid leukemia cells. The exposure to ELF-MF did not affect growth and viability of untreated K562 cells and did not influence the anti-proliferative effects of resveratrol, vinblastine, and etoposide. On the contrary, in quercetin-treated cells, exposure to ELF-MF significantly reduced the percentage of apoptotic cells and the caspase-3 activity and modified the cell cycle profile especially after 48xa0h of exposure. In addition, the simultaneous treatments for 24xa0h with quercetin plus ELF-MF increased Bcl-2 protein expression and prevented quercetin-induced downregulation of Mcl-1 and Bcl-xL. Finally, an increase of HSP70 expression was also observed after prolonged ELF-MF treatment. The ELF-MF-dependent modulation of the expression of anti-apoptotic Bcl-2 family and Hsp70 proteins could act as a pro-survival mechanism in K562 cells.