EMAIL THIS PAGE TO A FRIEND
Cold Spring Harbor protocols
Purification of dorsal root ganglion neurons from rat by immunopanning.
PMID 25086011
Abstract
Dorsal root ganglion neurons (DRGs) are sensory neurons that facilitate somatosensation and have been used to study neurite outgrowth, regeneration, and degeneration and PNS and CNS myelination. Studies of DRGs have relied on cell isolation strategies that generally involve extended culture in the presence of antimitotic agents or other cytotoxic treatments that target dividing cells. The surviving cells typically are dependent on serum for growth. Other methods, involving purification of DRGs based on their large size, produce low yield. In contrast, the immunopanning-based method described here for prospective isolation of DRGs from rodents allows for rapid purification in the absence of antimitotic agents and serum. These DRG cultures take place in a defined medium. They are free of Schwann cells and other glia and thus can be used to study the role of glia in the biology of DRG neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7880
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
C3H7NO2S · HCl · H2O