EMAIL THIS PAGE TO A FRIEND
Research in microbiology
Physiological features of Halomonas lionensis sp. nov., a novel bacterium isolated from a Mediterranean Sea sediment.
PMID 25086262
Abstract
A novel halophilic bacterium, strain RHS90(T), was isolated from marine sediments from the Gulf of Lions, in the Mediterranean Sea. Its metabolic and physiological characteristics were examined under various cultural conditions, including exposure to stressful ones (oligotrophy, high pressure and high concentrations of metals). Based on phylogenetic analysis of the 16S rRNA gene, the strain was found to belong to the genus Halomonas in the class Gammaproteobacteria. Its closest relatives are Halomonas axialensis and Halomonas meridiana (98% similarity). DNA-DNA hybridizations indicated that the novel isolate is genotypically distinct from these species. The DNA Gxa0+xa0C content of the strain is 54.4xa0mol%. The main fatty acids (C18:1ω7c, 2-OH iso-C15:0, C16:0 and/or C19:0 cyclo ω8c), main polar lipids (diphosphatidylglycerol, phosphatidylglycerol, phosphatidylethanolamine, phosphatidylcholine and an unidentified phosphoglycolipid) and major respiratory quinone (ubiquinone Q9) were determined. The novel isolate is heterotrophic, mesophilic, euryhaline (growth optimum ranging from 2 to 8% w/v NaCl) and is able to grow under stressful conditions. The strain accumulates poly-β-hydroxyalkanoates granules and compatible solutes. Based on genotypic, chemotaxonomic and phenotypic distinctiveness, this isolate is likely to represent a novel species, for which the name Halomonas lionensis is proposed. The type strain of H. lionensis is RHS90(T) (DSM 25632(T)xa0=xa0CIP 110370(T)xa0=xa0UBOCC 3186(T)).
Related Materials