EMAIL THIS PAGE TO A FRIEND
Basic research in cardiology
Cardiac fibroblasts support cardiac inflammation in heart failure.
PMID 25086637
Abstract
Cardiac remodeling and inflammation are hallmarks of cardiac failure and correlate with outcome in patients. However, the basis for the development of both remains unclear. We have previously reported that cardiac inflammation triggers transdifferentiation of fibroblasts to myofibroblasts and therefore increase accumulation of cardiac collagen, one key pathology in cardiac remodeling. Hence, identifying key pathways for inflammation would be beneficial for patients suffering from heart failure also. Besides their well-characterized function in matrix regulation, we here investigate the role of fibroblasts in the inflammatory process. We address for the first time the role of fibroblasts as inflammatory supporter cells in heart failure. Using endomyocardial biopsies from patients with heart failure and dilated cardiomyopathy, we created a primary human cardiac fibroblast cell culture system. We found that mechanical stretch mimicking cardiac dilation in heart failure induces activation of fibroblasts and not only stimulates production of extracellular matrix but more interestingly up-regulates chemokine production and triggers typical inflammatory pathways in vitro. Moreover, the cell culture supernatant of stretched fibroblasts activates inflammatory cells and induces further recruitment of monocytes by allowing transendothelial migration into the cardiac tissue. Our findings reveal that cardiac fibroblasts provide pro-inflammatory mediators and may act as sentinel cells activated by mechanical stress. Those cells are able to recruit inflammatory cells into the cardiac tissue, a process known to aggravate outcome of patients. This might be important in different forms of heart failure and therefore may be one general mechanism specific for fibroblasts.