EMAIL THIS PAGE TO A FRIEND
Ecotoxicology and environmental safety
Lead (Pb) in sheep exposed to mining pollution: implications for animal and human health.
PMID 25086824
Abstract
Livestock from the ancient mining area of Sierra Madrona and Alcudia Valley (Spain) is exposed to elevated levels of lead (Pb), as previous studies based on blood monitoring have revealed. Here we have studied blood, liver and muscle Pb levels in sheep in order to know if Pb exposure could represent a risk for human consumers of the meat and offal of these animals. A cross-sectional study was conducted with ≥4 years old (adults) ewes from the mining area (n=46) and a control area (n=21). Blood samples were taken before the sacrifice at the slaughterhouse, and liver and muscle samples were taken thereafter. At the same time, 2-3 year old rams (subadults, n=17) were blood sampled in the mining area. Blood, liver and muscle Pb levels were higher in the mining than in the control area. Blood Pb concentration in the mining area (n= 44, mean: 6.7μg/dl in ewes and 10.9μg/dl in rams) was above background levels (>6μg/dl) in 73.3 percent of animals. Liver Pb concentration in 68 percent of sheep from the mining area (n=32, mean: 6.16μg/g dry weight, d.w.) exceeded the minimum level associated with toxic exposure (5µg/g d.w.) and 87.5 percent of liver samples were above European Union Maximum Residue Levels (MRL) established for offal destined for human consumption (0.5µg/g w.w.~1.4µg/g d.w.). On the contrary, none of the muscle samples in ewes exceeded the EU MRL (0.1µg/g w.w.~0.34µg/g d.w.) established for meat, which may be related to the decline of blood Pb levels with age observed in the present study. These results suggest a potential health effect for sheep exposed to Pb pollution in this area and implications for food safety, but further research with lamb meat may be necessary to refine the risk assessment for human consumers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF32781999
Lead, microfoil, disks, 10mm, thinness 0.001μm, specific density 1.135μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF64961163
Lead, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.9μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF13065218
Lead, microfoil, disks, 10mm, thinness 0.1μm, specific density 103.4μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF47460753
Lead, microfoil, disks, 10mm, thinness 0.25μm, specific density 266.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF89832033
Lead, microfoil, disks, 10mm, thinness 1.0μm, specific density 1047.8μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF25514670
Lead, microfoil, disks, 25mm, thinness 1.0μm, specific density 1047.8μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF10093362
Lead, rod, 1000mm, diameter 12.7mm, 99.95%
Pb
GF79893444
Lead, rod, 1000mm, diameter 19.0mm, 99.95%
Pb
GF92060489
Lead, rod, 1000mm, diameter 3.2mm, 99.95%
Pb
GF66906197
Lead, rod, 1000mm, diameter 6.35mm, 99.95%
Pb
GF05266672
Lead, rod, 100mm, diameter 19.0mm, 99.95%
Pb
GF55722359
Lead, rod, 100mm, diameter 50mm, 99.95%
Pb
GF07560435
Lead, rod, 100mm, diameter 6.35mm, 99.95%
Pb
GF79027847
Lead, rod, 200mm, diameter 12.7mm, 99.95%
Pb
GF58334711
Lead, rod, 200mm, diameter 19.0mm, 99.95%
Pb
GF45053883
Lead, rod, 200mm, diameter 6.35mm, 99.95%
Pb
GF91839145
Lead, rod, 500mm, diameter 19.0mm, 99.95%
Pb
GF67936483
Lead, rod, 500mm, diameter 3.2mm, 99.95%
Pb
GF31735245
Lead, rod, 500mm, diameter 6.35mm, 99.95%
Pb
GF65793482
Lead, rod, 50mm, diameter 50mm, 99.95%
Pb
GF52882533
Lead, tube, 100mm, outside diameter 6.78mm, inside diameter 4.7mm, wall thickness 1.04mm, 99.9%
Pb
GF10316742
Lead, tube, 200mm, outside diameter 6.78mm, inside diameter 4.7mm, wall thickness 1.04mm, 99.9%
Pb
GF74266061
Lead, microfoil, disks, 25mm, thinness 0.25μm, specific density 266.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF86232225
Lead, rod, 100mm, diameter 12.7mm, 99.95%
Pb
GF99153085
Lead, rod, 200mm, diameter 50mm, 99.95%
Pb
GF99066190
Lead, tube, 1000mm, outside diameter 6.78mm, inside diameter 4.7mm, wall thickness 1.04mm, 99.9%
Pb
GF06993412
Lead, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.99+%
Pb
GF97912985
Lead, wire reel, 0.5m, diameter 2.0mm, as drawn, 99.99+%
Pb
GF66008167
Lead, wire reel, 10m, diameter 0.25mm, as drawn, 99.95%
Pb
GF31296551
Lead, wire reel, 1m, diameter 0.25mm, as drawn, 99.95%
Pb