EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
Calreticulin is localized at mitochondria of rat cardiomyocytes and affected by furazolidone.
PMID 25087122
Abstract
Calreticulin (CRT) is a calcium-buffering protein which is predominantly located in endoplasmic reticulum. In the previous mitochondria proteome analysis, we accidentally found that CRT may be also localized at myocardial mitochondria and was upregulated in a rat model of furazolidone-induced dilated cardiomyopathy. To our knowledge, there has not yet been any report of its presence in mitochondria of any cell types. The present study aimed to determine whether CRT was located at the mitochondria of rat cardiomyocytes and whether the mitochondrial CRT was affected by furazolidone. Mitochondrial preparations were isolated from primary cultured neonatal rat cardiomyocytes and purified by differential centrifugation. The purity of mitochondria was assessed by the reduction or elimination of the immunoreactivities of markers for cytosol, nucleus, sarcolemma, and endoplasmic reticulum. Western blot analysis demonstrated the presence of CRT in purified mitochondria of rat cardiomyocytes. The distribution of CRT to mitochondria was further confirmed by immuno-electron microscopy, flow cytometry, and laser scanning confocal microscopy (double staining with MitoTracker Red and CRT-Alexa Fluor 488). Western blot analysis also demonstrated that the mitochondrial content of CRT was significantly enhanced by furazolidone treatment by 2.73xa0±xa00.13 fold (Pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1401044
Anti-CALCR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401045
Anti-CALCR antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA028962
Anti-CALCR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV30114
Anti-CALR (AB1) antibody produced in rabbit, affinity isolated antibody
AV30113
Anti-CALR (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA002242
Anti-CALR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22432F Anti-Calreticulin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
C4606
Anti-Calreticulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2500268
Anti-COX4I1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405644
Anti-COX4I1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701802
Anti-cox4i1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA002485
Anti-COX4I1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501990
Anti-SLC6A8 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA008802
Anti-SLC6A8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV42248
Anti-SLC6A8 antibody produced in rabbit, IgG fraction of antiserum
SAB2104767
Anti-SLC6A8 antibody produced in rabbit, affinity isolated antibody
F9505
Furazolidone
C8H7N3O5
46297
Furazolidone, VETRANAL, analytical standard
C8H7N3O5
WH0000811M1
Monoclonal Anti-CALR antibody produced in mouse, clone 1G11-1A9, purified immunoglobulin, buffered aqueous solution
SAB5300527 Monoclonal Anti-CALR antibody produced in mouse, clone 1G6A7, ascites fluid
C7492
Monoclonal Anti-Calreticulin antibody produced in mouse, clone TO-11, tissue culture supernatant, buffered aqueous solution
SAB5300321
Monoclonal Anti-COX4I1 antibody produced in mouse, clone 6B3, ascites fluid
SAB1402152
Monoclonal Anti-COX4I1 antibody produced in mouse, clone 4A10, purified immunoglobulin, buffered aqueous solution