EMAIL THIS PAGE TO A FRIEND
The Journal of nutritional biochemistry
Maternal 18:3n-3 favors piglet intestinal passage of LPS and promotes intestinal anti-inflammatory response to this bacterial ligand.
PMID 25087993
Abstract
We recently observed that maternal 18:3n-3 increases piglet jejunal permeability. We hypothesized that this would favor intestinal lipopolysaccharide (LPS) passage and alter gut immune system education toward this bacterial ligand. Sows were fed 18:3n-3 or 18:2n-6 diets throughout gestation and lactation. In each litter, two piglets were given oral Gram-negative spectrum antibiotic from post-natal day (PND) 14 to 28. All piglets were weaned on a regular diet at PND28. 18:3n-3 piglets exhibited greater jejunal permeability to FITC-LPS at PND28. Levels of 18:3n-3 but neither 20:5n-3 nor 20:4n-6 were greater in mesenteric lymph nodes (MLN) of 18:3n-3 piglets. Jejunal explant or MLN cell cytokine responses to LPS were not influenced by the maternal diet. Antibiotic increased jejunal permeability to FITC-LPS and lowered the level of 20:5n-3 in MLN, irrespective of the maternal diet. At PND52, no long-lasting effect of the maternal diet or antibiotic treatment on jejunal permeability was noticed. 18:3n-3 and 20:4n-6 levels were greater and lower, respectively, in MLN of 18:3n-3 compared to 18:2n-6 piglets. IL-10 production by MLN cells in response to LPS was greater in the 18:3n-3 group, irrespective of the neonatal antibiotic treatment. IL-8 secretion by jejunal explants in response to LPS was lower in antibiotic-treated 18:3n-3 compared to 18:2n-6 piglets. Finally, proportion of MHC class II(+) antigen-presenting cells was greater in 18:3n-3 than 18:2n-6 MLN cells. In conclusion, maternal 18:3n-3 directs the intestinal immune response to LPS toward an anti-inflammatory profile beyond the breastfeeding period; microbiota involvement seems dependent of the immune cells considered.