EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Allelochemicals in the rhizosphere soil of Euphorbia himalayensis.
PMID 25088250
Abstract
Weed infestation has been known to cause considerable reductions in crop yields, thereby hindering sustainable agriculture. Many plants in genus Euphorbia affect neighboring plants and other organisms by releasing chemicals into the environment. In view of the serious threat of weeds to agriculture, the allelochemicals of Euphorbia himalayensis and their allelopathic effects were investigated. The extract of root exudates from rhizosphere soil exhibited allelopathic activities against crops (wheat, rape, and lettuce) and grasses (Poa annua, Festuca rubra, and red clover). Bioassay-guided fractionation and isolation from the root extract of E. himalayensis led to the characterization of two ellagic acid derivatives and a jatrophane diterpene, which observably showed phytotoxic activities against lettuce, Festuca arundinacea, and F. rubra. They were further confirmed by ultra-performance liquid chromatography-tandem mass spectrometry to have concentrations of 3.6, 3.8, and 8.99 nmol/g in the rhizospere soil, respectively. Bioassay indicated that the combination of the allelochemicals could be selective plant growth regulator in agriculture.