EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
Histone methyltransferase EZH2 is transcriptionally induced by estradiol as well as estrogenic endocrine disruptors bisphenol-A and diethylstilbestrol.
PMID 25088689
Abstract
Enhancer of Zeste homolog 2 (EZH2), a methyltransferase specific to histone 3 lysine 27, is a critical player in gene silencing and is overexpressed in breast cancer. Our studies demonstrate that EZH2 is transcriptionally induced by estradiol in cultured breast cancer cells and in the mammary glands of ovariectomized rats. EZH2 promoter contains multiple functional estrogen-response elements. Estrogen receptors (ERs) and ER coregulators such as mixed lineage leukemia (MLL) histone methylases (MLL2 and MLL3) and histone acetyltransferase CBP/P300 bind to the EZH2 promoter in the presence of estradiol and regulate estradiol-induced EZH2 expression. EZH2 expression is also increased upon exposure to estrogenic endocrine disrupting chemicals (EDCs) such as bisphenol-A (BPA) and diethylstilbestrol (DES). Similar to estradiol, BPA and DES-induced EZH2 expression is coordinated by ERs, MLLs and CBP/P300. In summary, we demonstrate that EZH2 is transcriptionally regulated by estradiol in vitro and in vivo, and its expression is potentially dysregulated upon exposure to estrogenic EDCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1500000
Diethylstilbestrol, European Pharmacopoeia (EP) Reference Standard
C18H20O2