EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Novel acylureidoindolin-2-one derivatives as dual Aurora B/FLT3 inhibitors for the treatment of acute myeloid leukemia.
PMID 25089810
Abstract
A series of 6-acylureido derivatives containing a 3-(pyrrol-2-ylmethylidene)indolin-2-one scaffold were synthesized as potential dual Aurora B/FLT3 inhibitors by replacing the 6-arylureido moiety in 6-arylureidoindolin-2-one-based multi-kinase inhibitors. (Z)-N-(2-(pyrrolidin-1-yl)ethyl)-5-((6-(3-(2-fluoro-4-methoxybenzoyl)ureido)-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide (54) was identified as a dual Aurora B/FLT3 inhibitor (IC50xa0=xa00.4xa0nM and 0.5xa0nM, respectively). Compound 54 also exhibited potent cytotoxicity with single-digit nanomolar IC50 values against the FLT3 mutant-associated human acute myeloid leukemia (AML) cell lines MV4-11 (FLT3-ITD) and MOLM-13 (FLT3-ITD). Compound 54 also specifically induced extrinsic apoptosis by inhibiting the phosphorylation of the Aurora B and FLT3 pathways in MOLM-13 cells. Compound 54 had a moderate pharmacokinetic profile. The mesylate salt of 54 efficiently inhibited tumor growth and reduced the mortality of BALB/c nude mice (subcutaneous xenograft model) that had been implanted with AML MOLM-13 cells. Compound 54 is more potent than sunitinib not only against FLT3-WT AML cells but also active against sunitinib-resistant FLT3-ITD AML cells. This study demonstrates the significance of dual Aurora B/FLT3 inhibitors for the development of potential agents to treat AML.