EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Cordycepin induces S phase arrest and apoptosis in human gallbladder cancer cells.
PMID 25090123
Abstract
Gallbladder cancer is the most common malignant tumor of the biliary tract, and this condition has a rather dismal prognosis, with an extremely low five-year survival rate. To improve the outcome of unresectable and recurrent gallbladder cancer, it is necessary to develop new effective treatments and drugs. The purpose of the present study was to evaluate the effects of cordycepin on human gallbladder cells and uncover the molecular mechanisms responsible for these effects. The Cell Counting Kit-8 (CCK-8) and colony formation assays revealed that cordycepin affected the viability and proliferation of human gallbladder cancer cells in a dose- and time-dependent manner. Flow cytometric analysis showed that cordycepin induced S phase arrest in human gallbladder cancer cell lines(NOZ and GBC-SD cells). Cordycepin-induced apoptosis was observed using an Annexin V/propidium iodide (PI) double-staining assay, and the mitochondrial membrane potential (ΔΨm) decreased in a dose-dependent manner. Additionally, western blot analysis revealed the upregulation of cleaved-caspase-3, cleaved-caspase-9, cleaved-PARP and Bax and the downregulation of Bcl-2, cyclin A and Cdk-2 in cordycepin-treated cells. Moreover, cordycepin inhibited tumor growth in nude mice bearing NOZ tumors. Our results indicate that this drug may represent an effective treatment for gallbladder carcinoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3428 Anti-Bax antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500343
Anti-Bax antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA027878
Anti-BAX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502549
Anti-BAX antibody produced in rabbit, affinity isolated antibody
B8429
Anti-Bax antibody, Mouse monoclonal, clone 6A7, purified from hybridoma cell culture
B8554
Anti-Bax antibody, Mouse monoclonal, clone 2D2, purified from hybridoma cell culture
PRS3335
Anti-Bcl-2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3337
Anti-Bcl-2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
B9804 Anti-Bcl-2 antibody, Mouse monoclonal, clone 10C4, purified from hybridoma cell culture
SAB4300339
Anti-BCL2 (Ab-56) antibody produced in rabbit, affinity isolated antibody
SAB4300340
Anti-BCL2 (Ab-70) antibody produced in rabbit, affinity isolated antibody
SAB2500154
Anti-BCL2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV01005
Anti-BCL2 antibody produced in rabbit, affinity isolated antibody
HPA001473
Anti-CASP9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503332
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503333
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503334
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503335
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
C7729 Anti-Caspase 9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300683
Anti-Caspase 9 antibody produced in rabbit, affinity isolated antibody
SAB3500405
Anti-Caspase-9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV01008 Anti-CDK2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV03006
Anti-CDK2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
C5223 Anti-Cdk2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous glycerol solution
SAB4503706
Anti-CDK2 antibody produced in rabbit, affinity isolated antibody
SAB2101602
Anti-NKX3-1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101603
Anti-NKX3-1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1412056
ANTI-NKX3-1 antibody produced in mouse, clone 3C1, purified immunoglobulin, buffered aqueous solution
SAB4500487
Anti-PARP (Cleaved-Asp214) antibody produced in rabbit, affinity isolated antibody
SAB4500488
Anti-PARP (Cleaved-Gly215) antibody produced in rabbit, affinity isolated antibody