EMAIL THIS PAGE TO A FRIEND
The Journal of steroid biochemistry and molecular biology
The vitamin D system is deregulated in pancreatic diseases.
PMID 25090635
Abstract
The vitamin D system is deregulated during development and progression of several cancer types. Data on the expression of the vitamin D system in the diseased pancreas are missing. The aim of this study was to investigate the expression of the vitamin D receptor (VDR), 1,25-dihydroxyvitamin D3 24-hydroxylase (CYP24A1), and the calcium-sensing receptor (CaSR), a vitamin D target gene, in the different regions of the pancreas in patients with chronic pancreatitis (n=6) and pancreatic ductal adenocarcinomas (PDAC) (n=17). We analyzed the expression of these genes at mRNA and protein level with quantitative real-time RT-PCR and immunostaining. mRNA expression of CYP24A1 and VDR was significantly increased in tumors compared with the adjacent non-tumorous tissue (p<0.01), while CaSR mRNA expression decreased. Both the VDR and the CaSR protein were highly expressed in the endocrine compared with the exocrine pancreas. In CP the CYP24A1 expression was highest in the endocrine pancreas, while in PDACs in the transformed ducts. In the PDAC patients CYP24A1 expression in the islets was significantly lower than in CP patients. Our data suggest that during ductal adenocarcinoma development the vitamin D system in the pancreas becomes deregulated on two levels: in the islets CYP24A1 expression decreases weakening the negative feedback regulation of the vitamin D-dependent insulin synthesis/secretion. In the transformed ducts CYP24A1 expression increases, impairing the antiproliferative effect of vitamin D in these cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA007269
Anti-SCTR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502719
Anti-SCTR antibody produced in rabbit, affinity isolated antibody
SAB1406459
Anti-SNCG antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA014404
Anti-SNCG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411301
Anti-SNCG antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2102673
Anti-VDR (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2102674
Anti-VDR (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2501098
Anti-VDR antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406580
Anti-VDR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2104917
Anti-VDR, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4503071
Anti-Vitamin D Receptor antibody produced in rabbit, affinity isolated antibody
SAB1402906
Monoclonal Anti-SCTR antibody produced in mouse, clone 3H1, purified immunoglobulin, buffered aqueous solution
WH0006623M1
Monoclonal Anti-SNCG antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB5300370
Monoclonal Anti-SNCG antibody produced in mouse, clone 1H10D2, ascites fluid
WH0007421M2
Monoclonal Anti-VDR antibody produced in mouse, clone 2F4, purified immunoglobulin, buffered aqueous solution