EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience research
Extracellular signal-related kinase 2/specificity protein 1/specificity protein 3/repressor element-1 silencing transcription factor pathway is involved in Aroclor 1254-induced toxicity in SH-SY5Y neuronal cells.
PMID 25093670
Abstract
Polychlorinated biphenyls (PCBs) cause a wide spectrum of toxic effects in the brain through undefined mechanisms. Exposure to the PCB mixture Aroclor-1254 (A1254) increases the repressor element-1 silencing transcription factor (REST) expression, leading to neuronal death. This study sought to understand the sequence of some molecular mechanisms to determine whether A1254 could increase REST expression and the cytoprotective effect of the phorbol ester tetradecanoylphorbol acetate (TPA) on A1254-induced toxicity in SH-SY5Y cells. As shown by Western blot analysis, A1254 (10 µg/ml) downregulates extracellular signal-related kinase 2 (ERK2) phosphorylation in a time-dependent manner, thereby triggering the binding of specificity protein 1 (Sp1) and Sp3 to the REST gene promoter as revealed by chromatin immunoprecipitation analysis. This chain of events results in an increase in REST mRNA and cell death, as assessed by quantitative real-time polymerase chain reaction and dimethylthiazolyl-2-5-diphenyltetrazolium-bromide assay, respectively. Accordingly, TPA prevented both the A1254-induced decrease in ERK2 phosphorylation and the A1254-induced increase in Sp1, Sp3, and REST protein expression. After 48 hr, TPA prevented A1254-induced cell death. ERK2 overexpression counteracted the A1254-induced increase in Sp1 and Sp3 protein expression and prevented A1254-induced Sp1 and Sp3 binding to the REST gene promoter, thus counteracting the increase in REST mRNA expression induced by the toxicant. In neuroblastoma SH-SY5Y cells, ERK2/Sp1/SP3/REST is a new pathway underlying the neurotoxic effect of PCB. The ERK2/Sp1/Sp3/REST pathway, which underlies A1254-induced neuronal death, might represent a new drug signaling cascade in PCB-induced neuronal toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV37192
Anti-SP1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV37193
Anti-SP1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2500981
Anti-SP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412220
ANTI-SP1 antibody produced in mouse, clone 4H6, purified immunoglobulin, buffered aqueous solution
SAB1412261
ANTI-SP1 antibody produced in mouse, clone 4B11, purified immunoglobulin, buffered aqueous solution
SAB1412262
ANTI-SP1 antibody produced in mouse, clone 2G5, purified immunoglobulin, buffered aqueous solution
SAB1412263
ANTI-SP1 antibody produced in mouse, clone 3H7, purified immunoglobulin, buffered aqueous solution
SAB1412264
ANTI-SP1 antibody produced in mouse, clone 1G9, purified immunoglobulin, buffered aqueous solution
S9809 Anti-Sp1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001853
Anti-SP1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV43585
Anti-SP1 antibody produced in rabbit, affinity isolated antibody
HPA012292
Anti-SP1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502836
Anti-SP1 antibody produced in rabbit, affinity isolated antibody
SAB2103396
Anti-SP3 antibody produced in rabbit, affinity isolated antibody
PLA0307
Goat anti-Sp1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
WH0006667M2
Monoclonal Anti-SP1 antibody produced in mouse, clone 1A5, purified immunoglobulin, buffered aqueous solution
SAB1404397
Monoclonal Anti-SP1 antibody produced in mouse, clone 4C8, purified immunoglobulin, buffered aqueous solution
PLA0044
Rabbit anti-Sp1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.