EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Critical role of all-trans retinoic acid in stabilizing human natural regulatory T cells under inflammatory conditions.
PMID 25099355
Abstract
Recent studies have demonstrated that thymus-derived naturally occurring CD4(+)Foxp3(+) regulatory T cells (Tregs) in human and mouse may be unstable and dysfunctional in the presence of proinflammatory cytokines. All-trans RA (atRA), the active derivative of vitamin A, has been shown to regulate Treg and T effector cell differentiation. We hypothesize atRA stabilizes human natural Tregs (nTregs) under inflammatory conditions. atRA prevents human nTregs from converting to Th1 and/or Th17 cells and sustains their Foxp3 expression and suppressive function in vitro or in vivo following encounters with IL-1 and IL-6. Interestingly, adoptive transfer of human nTregs pretreated with atRA significantly enhanced their suppressive effects on xenograft-vs.-host diseases (xGVHDs), and atRA- but not rapamycin-pretreated nTregs sustained the functional activity against xGVHD after stimulation with IL-1/IL-6. atRA suppresses IL-1 receptor (IL-1R) up-regulation, accelerates IL-6R down-regulation, and diminishes their signaling events as well as prevents the up-regulation of STIP1 homology and U-Box containing protein 1 on Foxp3(+) cells following IL-1/IL-6 stimulation. atRA also increases histone acetylation on Foxp3 gene promoter and CpG demethylation in the region of Foxp3 locus (i.e., Treg-specific demethylated region). These results strongly implicate that nTregs primed with atRA may represent a novel treatment strategy to control established chronic immune-mediated autoimmune and inflammatory diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1300799
Anti-α-Synuclein (N-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
S3062
Anti-α-Synuclein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
S5566
Anti-α-Synuclein antibody, Mouse monoclonal, clone Syn211, purified from hybridoma cell culture
SAB1406018
Anti-IL1R1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
PRS4065
Anti-PD-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4067
Anti-PD-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900280
Anti-PDCD1 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900373
Anti-PDCD1 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2500763
Anti-PDCD1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400196
Anti-PDCD1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA035981
Anti-PDCD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300502
Anti-SNCA (Ab-125) antibody produced in rabbit, affinity isolated antibody
SAB4300445
Anti-SNCA (Ab-129) antibody produced in rabbit, affinity isolated antibody
SAB4300528
Anti-SNCA (Ab-133) antibody produced in rabbit, affinity isolated antibody
SAB2502111
Anti-SNCA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502112
Anti-SNCA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102253
Anti-SNCA antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA005459
Anti-SNCA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1105828
Anti-SPATA2 (341-354) antibody produced in rabbit, IgG fraction of antiserum
SAB1410026
Anti-SPATA2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA048581
Anti-SPATA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107365
Anti-SPATA2 antibody produced in rabbit, affinity isolated antibody
SAB1411697
Anti-SPATA2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1105169 Anti-STAT1 (671-685) antibody produced in rabbit, IgG fraction of antiserum
SAB4300326
Anti-STAT1 (Ab-701) antibody produced in rabbit, affinity isolated antibody
SAB4300441
Anti-STAT1 (Ab-727) antibody produced in rabbit, affinity isolated antibody
GW22758 Anti-STAT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV33373
Anti-STAT1 (AB1) antibody produced in rabbit, affinity isolated antibody
GW22780 Anti-STAT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV33374
Anti-STAT1 (AB2) antibody produced in rabbit, affinity isolated antibody