EMAIL THIS PAGE TO A FRIEND
Clinical otolaryngology : official journal of ENT-UK ; official journal of Netherlands Society for Oto-Rhino-Laryngology & Cervico-Facial Surgery
Effects of adenoid/tonsillectomy on inflammatory response in snoring children with witnessed apnoea.
PMID 25099745
Abstract
The aim of this study was to investigate the clinical significance of preoperative serum C-reactive protein (CRP), interleukin-6 (IL-6), fetuin-A, cystatin C, adiponectin and tumour necrosis factor-alpha (TNF-α) levels in children with adenoid/tonsillar hypertrophy and compare these results with postoperative values. Prospective clinical trial. Single tertiary care centre. Thirty-five children (14 boys and 21 girls) participated in this study. The operations were performed with the indication of sleep-disordered breathing (SDB). Blood samples were taken preoperatively and at 6xa0months postoperatively to determine the changes in serum high-sensitivity CRP (hs-CRP), IL-6, fetuin-A, cystatin C, adiponectin and TNF-α levels. Pre- and postoperative values of body weights and heights of the participants were recorded preoperatively and at postoperative 6xa0months. Patients underwent adenoidectomy (nxa0=xa021) or adenotonsillectomy (nxa0=xa021). The mean age at surgery was 8.74xa0±xa03.33xa0years (range 3-16xa0years). Mean serum levels of parameters measured preoperatively and at postoperative 6xa0months. The observed differences between pre- and postoperative values were statistically significant (Pxa0xa00.05). High levels of IL-6, hs-CRP and TNF-α and low levels of fetuin-A in serum might have a clinical significance in patients with sleep-disordered breathing. Levels of cytokines in children with sleep-disordered breathing because of adenotonsillar disease decreased after surgical treatment. The risks of development of cardiovascular disease are decreased in association with lower levels of cytokines, which are also closely associated with many disease states.
Related Materials