PloS one
Germinal center B cell depletion diminishes CD4+ follicular T helper cells in autoimmune mice.
PMID 25101629
Abstract
Continuous support from follicular CD4(+) T helper (Tfh) cells drives germinal center (GC) responses, which last for several weeks to produce high affinity memory B cells and plasma cells. In autoimmune Sle1 and NZB/W F1 mice, elevated numbers of Tfh cells persist, promoting the expansion of self-reactive B cells. Expansion of circulating Tfh like cells have also been described in several autoimmune diseases. Although, the signals required for Tfh differentiation have now been well described, the mechanisms that sustain the maintenance of fully differentiated Tfh are less understood. Recent data demonstrate a role for GC B cells for Tfh maintenance after protein immunization. Given the pathogenic role Tfh play in autoimmune disease, we explored whether B cells are required for maintenance of autoreactive Tfh. Our data suggest that the number of mature autoreactive Tfh cells is controlled by GC B cells. Depletion of B cells in Sle1 autoimmune mice leads to a dramatic reduction in Tfh cells. In NZB/W F1 autoimmune mice, similar to the SRBC immunization model, GC B cells support the maintenance of mature Tfh, which is dependent mainly on ICOS. The CD28-associated pathway is dispensable for Tfh maintenance in SRBC immunized mice, but is required in the spontaneous NZB/W F1 model. These data suggest that mature Tfh cells require signals from GC B cells to sustain their optimal numbers and function in both autoimmune and immunization models. Thus, immunotherapies targeting B cells in autoimmune disease may affect pathogenic Tfh cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1300799
Anti-α-Synuclein (N-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
S3062
Anti-α-Synuclein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
S5566
Anti-α-Synuclein antibody, Mouse monoclonal, clone Syn211, purified from hybridoma cell culture
SAB1408550
Anti-BLR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
C6487 Anti-CD40 Ligand antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2100377
Anti-CD40LG antibody produced in rabbit, affinity isolated antibody
HPA045827
Anti-CD40LG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA042455 Anti-CR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA043579
Anti-CR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA049348
Anti-CR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C3366 Anti-CXCR5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA042432
Anti-CXCR5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406012
Anti-IGHD antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001895
Anti-LAMB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1300714
Anti-SLAMF1 (Y307) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1406444
Anti-SLAMF1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB3500996
Anti-SLAMF1 antibody produced in rabbit, affinity isolated antibody
SAB4300502
Anti-SNCA (Ab-125) antibody produced in rabbit, affinity isolated antibody
SAB4300445
Anti-SNCA (Ab-129) antibody produced in rabbit, affinity isolated antibody
SAB4300528
Anti-SNCA (Ab-133) antibody produced in rabbit, affinity isolated antibody
SAB2502111
Anti-SNCA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502112
Anti-SNCA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102253
Anti-SNCA antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA005459
Anti-SNCA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1105828
Anti-SPATA2 (341-354) antibody produced in rabbit, IgG fraction of antiserum
SAB1410026
Anti-SPATA2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA048581
Anti-SPATA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107365
Anti-SPATA2 antibody produced in rabbit, affinity isolated antibody
SAB1411697
Anti-SPATA2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0000643M1
Monoclonal Anti-BLR1 antibody produced in mouse, clone 2C1, purified immunoglobulin, buffered aqueous solution