EMAIL THIS PAGE TO A FRIEND
Protein and peptide letters
Functional characterisation and permeation studies of lyophilised thiolated chitosan xerogels for buccal delivery of insulin.
PMID 25101633
Abstract
Stable and mucoadhesive, lyophilised, thiolated chitosan xerogels, loaded with insulin for buccal mucosa deliv- ery, in place of the currently used parenteral route have been developed. The xerogels were backed with impervious ethyl- cellulose laminate to ensure unidirectional release and also loaded with enzyme inhibitor to enhance insulin permeability across the buccal mucosa. Characterisation of xerogels using(1) HNMR confirmed the degree of deacetylation of the syn- thesised thiolated chitosan. The amount of thiol groups immobilised on the modified chitosan was quantified by Ellman's reaction and molecular weight monitored by gel permeation chromatography. The stability of the secondary structure of insulin was examined by attenuated total reflectance Fourier transform infra-red spectroscopy and circular dichroism. In vitro and ex vivo permeation studies were undertaken by using EpiOral ™ and sheep buccal membrane respectively. Insu- lin released from thiolated chitosan xerogels, loaded with aprotinin (enzyme inhibitor and permeation enhancer) showed a 1.7-fold increase in permeation through EpiOral ™ buccal tissue construct compared to the pure drug. However, permea- tion was decreased for xerogels containing the enzyme inhibitor glutathione. Further, aprotinin containing xerogels en- hanced insulin permeation through sheep buccal membrane and demonstrated good linear correlation with the permeation data from the EpiOral ™ study. The results show the potential application of lyoph ilised thiolated chitosan xerogels con- taining aprotinin with improved mucoadhesion, penetration enhancing and enzyme inhibition characteristics for buccal mucosa delivery of macromolecules such as insulin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7000
1-Naphthyl phosphate monosodium salt monohydrate, ≥98% (titration), powder
C10H8NaO4P · H2O
C9157
Activated charcoal, suitable for cell culture, suitable for plant cell culture
C
C6289
Activated charcoal, plant cell culture tested
C
390127
Activated charcoal, purified by resistive heating of graphite
C
05112
Activated charcoal, for GC
C
05105
Activated charcoal, powder
C
31616
Activated charcoal, puriss. p.a., powder
C
80485
Activated charcoal, for the determination of AOX, 50-150 μm particle size
C
440345
Glass spheres, 9-13 μm particle size
O2Si
420875
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
O2Si
420832
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420840
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420883
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
O2Si
420891
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
O2Si
420824
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420816
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420808
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
O2Si
420794
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
O2Si
420786
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420778
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
O2Si
420859
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
O2Si
83340
Sand, white quartz, purum p.a., powder
O2Si
204358
Sand, white quartz, ≥99.995% trace metals basis
O2Si
421553
Silica
O2Si
381276
Silica, 99.8%
O2Si
718483
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
O2Si
748161
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
O2Si
S5505
Silica, fumed, powder, 0.2-0.3 μm avg. part. size (aggregate)
O2Si
S5130
Silica, fumed, powder
O2Si
560979
Silica, mesostructured, MSU-F (cellular foam)
O2Si