EMAIL THIS PAGE TO A FRIEND
Platelets
Identification of anti-HPA-1a allo-antibodies using IgG platelet antibody detection and crossmatch system assay with Galileo Echo.
PMID 25101933
Abstract
Fetal/neonatal allo-immune thrombocytopenia is the most frequent and the most dangerous clinical condition involving anti-human platelet antigens (HPA)-1a allo-antibodies. Anti-HPA-1a allo-immunization requires rapid and accurate diagnosis to determine appropriate treatment. The Capture-P Ready-Screen assay (C-PRS) is a new qualitative immunoassay to detect IgG anti-human leukocyte antigen (HLA) and anti-HPA allo-antibodies. The aim of this study is to assess the identification of anti-HPA-1a allo-antibodies using the C-PRS assay, associated with HLA class I stripping reagents, on the automated benchtop analyzer Galileo Echo. Forty-nine sera were analyzed: without anti-HLA class I or anti-HPA allo-antibodies, with anti-HLA class I allo-antibodies, with anti-HPA-1a allo-antibodies, among which with anti-HLA class I allo-antibodies. None of the samples without allo-antibodies were reactive. Only anti-HLA antibodies, detected by cytotoxicity-dependent complement and not by Luminex, remained positive before and after stripping reagents. Of the 13 samples, anti-HPA-1a allo-antibodies that were correctly identified before and after incubation with HLA assassin reagent were 70% and 85%, respectively. Anti-glycoprotein auto-antibodies and anti-HLA allo-antibodies do not interfere with the detection of anti-HPA-1a antibodies. This preliminary study indicates that further improvement of the test will be helpful in developing a clinically useful assay in the future.