EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Solid supported in situ derivatization extraction of acidic degradation products of nerve agents from aqueous samples.
PMID 25103280
Abstract
This study deals with the solid supported in situ derivatization extraction of acidic degradation products of nerve agents present in aqueous samples. Target analytes were alkyl alkylphosphonic acids and alkylphosphonic acids, which are important environmental signatures of nerve agents. The method involved tert-butyldimethylchlorosilane mediated in situ silylation of analytes on commercially available diatomaceous solid phase extraction cartridges. Various parameters such as derivatizing reagent, its concentration, reaction time, temperature and eluting solvent were optimized. Recoveries of the analytes were determined by GC-MS which ranged from 60% to 86%. The limits of detection (LOD) and limit of quantification (LOQ) with selected analytes were achieved down to 78 and 213ngmL(-1) respectively, in selected ion monitoring mode. The successful applicability of method was also demonstrated on samples of biological origin such as plasma and to the samples received in 34th official proficiency test conducted by the Organization for Prohibition the of Chemical Weapons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
255327
Tripropyl phosphate, 99%
C9H21O4P