EMAIL THIS PAGE TO A FRIEND
Molecular cancer research : MCR
The nuclear pool of tetraspanin CD9 contributes to mitotic processes in human breast carcinoma.
PMID 25103498
Abstract
Tetraspanin-29 (CD9) is an integral membrane protein involved in several fundamental cell processes and in cancer metastasis. Here, characterization of a panel of breast cancer cells revealed a nuclear pool of CD9, not present in normal human mammary epithelial cells. Antibody binding to surface CD9 of breast cancer cells resulted in increased nuclear CD9 fluorescence. CD9 was also found, along with a plasma membrane-associated pool, in the nuclei of all primary ductal breast carcinoma patient specimens analyzed. In all patients, about 40% of the total CD9 cellular fluorescence was nuclear. CD9 colocalized at the nuclear level with CEP97, a protein implicated in centrosome function, and with the IGSF8, an established CD9 partner in the plasma membrane. Co-immunoprecipitation of CEP97 and IGSF8 with CD9 was shown in nuclear extracts from breast cancer cells expressing a CD9-GFP fusion protein. However, by fluorescence resonance energy transfer (FRET) analysis, no direct binding of CD9 with either protein was observed, suggesting that CD9 is part of a larger nuclear protein complex. CD9 depletion or exposure of parental breast cancer cells to anti-CD9 mAb resulted in polynucleation and multipolar mitoses. These data indicate that the nuclear CD9 pool has an important role in the mitotic process. The discovery of a nuclear pool of CD9 has prognostic and/or therapeutic potential for patients with ductal carcinoma of the breast.