EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Characterisation of triterpenes and new phenolic lipids in Cameroonian propolis.
PMID 25104230
Abstract
Chemical investigation of a sample of propolis originating from North-Western Cameroon led to the isolation of thirteen alk(en)ylphenols (1-13) (inseparable mixture) along with α-amyrin (14), β-amyrin (15), lupeol (16), cycloartenol (17), mangiferonic acid (18), ambonic acid (19), mangiferolic acid (20), ambolic acid (21), isomangiferolic acid (22) and nine alk(en)ylresorcinols (23-31) (inseparable mixture). All compounds were identified following analysis of their spectroscopic data and comparison with previously published reports. Compounds (8), (12), (13) and (30) are new natural products. GC-MS analysis carried out on the alk(en)ylphenol and alk(en)ylresorcinol mixtures (dimethyl disulphide trimethylsilyl derivatives) revealed the presence of saturated and mono-unsaturated compounds with side chain lengths ranging from C11 to C19 and C15 to C19, respectively. The position of the double bond in mono-unsaturated derivatives was established from the characteristic fragments resulting from the cleavage of the bond between the two methylthio-substituted carbons. The most abundant compound in each mixture was 3-(12'Z-heptadecenyl)-phenol (10) and 5-(12'Z-heptadecenyl)-resorcinol (29). This study is the first to report the presence of triterpenes (except for lupeol) and phenolic lipids, including eighteen compounds previously unreported in bee glue, in an African sample.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04016
3-Pentadecylphenol, analytical standard
C21H36O
P4402
3-Pentadecylphenol, technical grade, 90%
C21H36O
97001
5-Heptadecylresorcinol, analytical standard
C23H40O2
91822
5-Pentadecylresorcinol, analytical standard
C21H36O2
386529
Chlorotrimethylsilane, purified by redistillation, ≥99%
C3H9ClSi
95541
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
C3H9ClSi
92361
Chlorotrimethylsilane, ≥98.0% (GC)
C3H9ClSi
92360
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
C3H9ClSi
89595
Chlorotrimethylsilane, for GC derivatization, ≥99.0% (GC)
C3H9ClSi
385433
Chlorotrimethylsilane solution, 1.0 M in methylene chloride
C3H9ClSi
384410
Chlorotrimethylsilane solution, 1.0 M in THF
C3H9ClSi
08172
Cycloartenol, ≥90% (GC)
C30H50O
471569
Dimethyl disulfide, ≥99.0%
C2H6S2
320412
Dimethyl disulfide, 99%
C2H6S2
W353604
Dimethyl disulfide, ≥98%, FG
C2H6S2
68986
Dimethyl disulfide, analytical standard
C2H6S2
Y0000012
Hymecromone impurity A, European Pharmacopoeia (EP) Reference Standard
C6H6O2
447420
Orcinol, 97%
C7H8O2
97063
p-Anisaldehyde, analytical standard
C8H8O2
A88107
p-Anisaldehyde, 98%
C8H8O2
W267007
p-Anisaldehyde, ≥97.5%, FCC, FG
C8H8O2
W267020
p-Anisaldehyde, natural, 98%, FG
C8H8O2
53363
Resorcinol, certified reference material, TraceCERT®
C6H6O2
R5645
Resorcinol, BioXtra, ≥99%
C6H6O2
398047
Resorcinol, ACS reagent, ≥99.0%
C6H6O2
307521
Resorcinol, ReagentPlus®, 99%
C6H6O2
R406
Resorcinol, reagent grade, 98%
C6H6O2
16101
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
C6H6O2
83601
Resorcinol, tested according to Ph.Eur.
C6H6O2
W358908
Resorcinol, ≥98%, FG
C6H6O2