EMAIL THIS PAGE TO A FRIEND
Breast cancer research and treatment
Vaccination with ErbB-2 peptides prevents cancer stem cell expansion and suppresses the development of spontaneous tumors in MMTV-PyMT transgenic mice.
PMID 25104444
Abstract
ErbB-2 has been implicated as a target for cancer-initiating cells in breast and other cancers. ErbB-2-directed peptide vaccines have been shown to be effective in prevention of spontaneous tumorigenesis of breast in neu transgenic mouse model, and cellular immunity is proposed as a mechanism for the anti-tumor efficacy. However, there has been no explanation as to how immunity suppresses tumorigenesis from the early stage carcinogenesis, when ErbB-2 expression in breast is low. Here, we investigated a peptide-based vaccine, which consists of two MHC class II epitopes derived from murine ErbB-2, to prevent the occurrence of spontaneous tumors in breast and assess immune impact on breast cancer stem cells. Female MMTV-PyMT transgenic mice were immunized with either ErbB-2 peptide vaccine, or a peptide from tetanus toxoid, or PBS in immune adjuvant. ErbB-2 peptides vaccine completely suppressed spontaneous breast tumors, and the efficacy was correlated with antigen-specific T-cell and antibody responses. In addition, immune serum from the mice of ErbB-2 vaccine group had an inhibitory effect on mammosphere-forming capacity and signaling through ErbB-2 and downstream Akt pathway in ErbB-2 overexpressing mouse mammary cancer cells. We provide evidence that multi-epitope class II peptides vaccine suppresses tumorigenesis of breast potentially by inhibiting the growth of cancer stem cells. We also suggest that a strategy of inducing strong immune responses using multi-epitope ErbB-2-directed helper vaccine might be useful in preventing breast cancer recurrence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22757 Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22775 Anti-AKT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501934
Anti-AKT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
GW22768 Anti-MAPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA030069
Anti-MAPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503853
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
SAB4504016
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4504015
Anti-phospho-Akt (pSer246) antibody produced in rabbit, affinity isolated antibody
SAB4504332
Anti-phospho-Akt (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504017
Anti-phospho-Akt (pThr72) antibody produced in rabbit, affinity isolated antibody
SAB4504110
Anti-phospho-Akt (pTyr326) antibody produced in rabbit, affinity isolated antibody
SAB4504014
Anti-phospho-Akt (pTyr474) antibody produced in rabbit, affinity isolated antibody
SAB4300043
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504749
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4503852
Anti-phospho-Akt1 (pThr450) antibody produced in rabbit, affinity isolated antibody
SAB4300260
Anti-phospho-AKT1 (pThr450) antibody produced in rabbit, affinity isolated antibody
P3862 Anti-phospho-PKB (pThr308) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
WH0000207M3
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
SAB1400005
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6D6, purified immunoglobulin, buffered aqueous solution
SAB1400006
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6G6, purified immunoglobulin, buffered aqueous solution
SAB1400007
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2H1, purified immunoglobulin, buffered aqueous solution
SAB1400008
Monoclonal Anti-AKT1 antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution
SAB1305558 MONOCLONAL ANTI-ERK2 antibody produced in mouse, clone 814CT5.4.2, IgG fraction of antiserum, buffered aqueous solution
WH0005594M1
Monoclonal Anti-MAPK1 antibody produced in mouse, clone 1D1, purified immunoglobulin, buffered aqueous solution
R2650 Monoclonal Anti-Rac antibody produced in mouse, clone 23A8, purified immunoglobulin, buffered aqueous solution
PLA0236
Rabbit anti-ERK2 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.