EMAIL THIS PAGE TO A FRIEND
Virology
Attenuation of virus production at high multiplicities of infection in Aureococcus anophagefferens.
PMID 25104555
Abstract
Infection dynamics (saturation kinetics, infection efficiency, adsorption and burst size) for the Aureococcus anophagefferens-Brown Tide virus (AaV) system were investigated using susceptible and resistant strains. Adsorption assays revealed that virus affinity to the cell surface is a key determinant of infectivity. Saturation of infection occurred at a multiplicity of infection (MOI) of 8 viruses per host and resulted in ~90-95% of infected cells, with burst sizes ranging from 164 to 191. Insight from the AaV genome implicates recycling of host nucleotides rather than de novo synthesis as a constraint on viral replication. Viral yields and mean burst sizes were significantly diminished with increasing MOI. This phenomenon, which was reminiscent of phage-induced 'lysis from without', appeared to be caused by viral contact and was unrelated to bacteria, signaling/toxic compounds, or defective interfering viruses. We posit that high-MOI effects attenuate viral proliferation in natural systems providing a negative feedback on virus-induced bloom collapse.