EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
Theoretical and Experimental Studies of N,N-Dimethyl-N'-Picryl-4,4'-Stilbenediamine.
PMID 25106474
Abstract
N,N-dimethyl-N'-picryl-4,4'-stilbenediamine (DMPSDA) was prepared, purified and crystallised in a form of black lustrous crystals, and its absorption and fluorescence spectra were recorded in cyclohexane, acetonitrile and dimethyl sulfoxide. Non-emissive intramolecular charge transfer state (ICT) was clearly observed in this molecule in all three solvents. Theoretical calculations demonstrating a betaine electronic structure of the trinitrophenyl group in the ground state of the molecule and a charge transfer nature of the long wavelength transition S0 → S1 supported the experimental observations of the ICT formation in the molecule.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
156477
4-(Dimethylamino)benzaldehyde, ACS reagent, 99%
C9H11NO