EMAIL THIS PAGE TO A FRIEND
Stem cell research & therapy
Viability of equine mesenchymal stem cells during transport and implantation.
PMID 25107289
Abstract
Autologous mesenchymal stem cell (MSC) injection into naturally-occurring equine tendon injuries has been shown to be safe and efficacious and protocols inform translation of the technique into humans. Efficient transfer of cells from the laboratory into tissue requires well-validated transport and implantation techniques. Cell viability in a range of media was determined over 72xa0hours and after injection through a 19G, 21G or 23G needle. Viability, proliferation and apoptosis were analysed using TrypanBlue, alamarBlue® and AnnexinV assays. Cell viability was similar in all re-suspension media following 24xa0hour storage, however cell death was most rapid in bone marrow aspirate, platelet-rich plasma and serum after longer storage. Cryogenic media exhibited greatest viability regardless of storage time. Cell proliferation after 24 and 72xa0hour storage was similar for all media, except after 24xa0hours in serum wherein proliferation was enhanced. MSC tri-lineage differentiation and viability did not significantly change when extruded through 19G, 21G or 23G needles, but 21G and 23G needles significantly increased apoptotic cells compared to 19G and non-injected controls. All gauges induced a decrease in metabolic activity immediately post-injection but cells recovered by 2xa0hours. These data indicate storage and injection influence viability and subsequent cell behaviour and provide recommendations for MSC therapy that implantation of cells should occur within 24xa0hours of recovery from culture, using larger needle bores.