EMAIL THIS PAGE TO A FRIEND
International immunopharmacology
Analysis of cytokine release assay data using machine learning approaches.
PMID 25107440
Abstract
The possible onset of Cytokine Release Syndrome (CRS) is an important consideration in the development of monoclonal antibody (mAb) therapeutics. In this study, several machine learning approaches are used to analyze CRS data. The analyzed data come from a human blood in vitro assay which was used to assess the potential of mAb-based therapeutics to produce cytokine release similar to that induced by Anti-CD28 superagonistic (Anti-CD28 SA) mAbs. The data contain 7 mAbs and two negative controls, a total of 423 samples coming from 44 donors. Three (3) machine learning approaches were applied in combination to observations obtained from that assay, namely (i) Hierarchical Cluster Analysis (HCA); (ii) Principal Component Analysis (PCA) followed by K-means clustering; and (iii) Decision Tree Classification (DTC). All three approaches were able to identify the treatment that caused the most severe cytokine response. HCA was able to provide information about the expected number of clusters in the data. PCA coupled with K-means clustering allowed classification of treatments sample by sample, and visualizing clusters of treatments. DTC models showed the relative importance of various cytokines such as IFN-γ, TNF-α and IL-10 to CRS. The use of these approaches in tandem provides better selection of parameters for one method based on outcomes from another, and an overall improved analysis of the data through complementary approaches. Moreover, the DTC analysis showed in addition that IL-17 may be correlated with CRS reactions, although this correlation has not yet been corroborated in the literature.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22573F Anti-BB1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA024353
Anti-CD22 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300687
Anti-CD22 antibody produced in rabbit, affinity isolated antibody
SAB2501381
Anti-Cd28 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501674
Anti-CD28 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405583
Anti-CD28 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
C7831 Anti-CD28 antibody, Mouse monoclonal, clone CD28.2, purified from hybridoma cell culture
SAB2501656
Anti-CD80 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405584
Anti-CD80 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101153
Anti-IL5 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2107055
Anti-IL5 antibody produced in rabbit, affinity isolated antibody
I9898 Anti-Interleukin-5 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
SAB4700126 Monoclonal Anti-CD22 antibody produced in mouse, clone MEM-01, purified immunoglobulin, buffered aqueous solution
SAB4700135 Monoclonal Anti-CD28 antibody produced in mouse, low endotoxin, clone CD28.2, purified immunoglobulin, buffered aqueous solution
SAB4700739 Monoclonal Anti-CD28 antibody produced in mouse, clone JJ319, purified immunoglobulin, buffered aqueous solution
SAB4700751
Monoclonal Anti-CD28 antibody produced in mouse, clone CD28.2, purified immunoglobulin, buffered aqueous solution
WH0000941M1 Monoclonal Anti-CD80 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1403647
Monoclonal Anti-CD80 antibody produced in mouse, clone 3E10, ascites fluid
SAB4700140 Monoclonal Anti-CD80 antibody produced in mouse, clone MEM-233, purified immunoglobulin, buffered aqueous solution