EMAIL THIS PAGE TO A FRIEND
ACS nano
Thin films of homopolymers and cylinder-forming diblock copolymers under shear.
PMID 25107567
Abstract
We study thin films of homopolymers (PS) and monolayers of cylinder-forming diblock copolymers (PS–PHMA) under shear. To this end, we employed both experiments and computer simulations that correctly take into account hydrodynamic interactions and chain entanglements. Excellent quantitative agreement for static as well as dynamic properties in both the homopolymer and diblock copolymer cases was achieved. In particular, we found that the homopolymer thin films exhibit a distinct shear thinning behavior, which is strongly correlated with the disentanglement and shear alignment of the constituent polymer chains. For the PS–PHMA films, we show that shear can be employed to induce long-range ordering to the spontaneously self-assembled microdomains, which is required for many applications such as the fabrication of nanowire arrays. We found that the impact of chemical incompatibility on the viscosity is only minor in shear-aligned films. Once the domains were aligned, the films exhibited an almost Newtonian response to shear because the cylindrical microdomains acted as guide rails, along which the constituent copolymer chains could simply slide. Furthermore, we developed a model for predicting the onset of shear alignment based on equilibrium dynamics data, and found good agreement with our shear simulations. The employed computational method holds promise for a faster and more cost-effective route for developing custom tailored materials.