EMAIL THIS PAGE TO A FRIEND
Brain, behavior, and immunity
Acute psychosocial stress induces differential short-term changes in catecholamine sensitivity of stimulated inflammatory cytokine production.
PMID 25107875
Abstract
We have previously shown that psychosocial stress induces acute changes in glucocorticoid (GC) sensitivity of pro-inflammatory cytokine production. However, hormones of the sympathetic adrenal medullary system complement endocrine regulation of inflammatory responses. The current study therefore aimed at investigating the effects of repeated acute stress exposure on catecholamine sensitivity of inflammatory cytokine production. Twenty healthy male participants were subjected to the Trier Social Stress Test on two consecutive days. Blood samples were taken before and repeatedly after stress. Whole blood was stimulated with lipopolysaccharide and incubated with increasing concentrations of epinephrine (E) and norepinephrine (NE) for 18h. Tumor-necrosis-factor (TNF) alpha and interleukin (IL)-6 were measured in culture supernatants. Overall, incubation with E and NE induced dose-dependent suppression of TNF-alpha (NE: F=77.66, p<.001; E: F=63.38, p<.001), and IL-6 production (NE: F=28.79, p<.001; E: F=24.66, p<.001). Acute stress exposure resulted in reduced sensitivity of TNF-alpha (NE: F=6.36, p<.001; E: F=4.86, p=.005), but not IL-6 (NE: F=1.07, p=.38; E: F=0.88, p=.50) to the inhibitory signals of E and NE. No evidence of habituation of these effects was found (all p⩾.22). The present findings extend our knowledge on changes in inflammatory target tissue sensitivity in response to acute psychosocial stress from glucocorticoid-dependent effects to catecholamine-dependent effects. Stress-induced decreases in catecholamine sensitivity thereby suggest intracellular processes aiding in maintaining a healthy endocrine-immune interplay. Longitudinal studies will have to investigate the processes leading from a supposedly beneficial short-term catecholamine resistance in response to acute stress to basal catecholamine resistance observed in relation to negative health outcomes.