EMAIL THIS PAGE TO A FRIEND
Experimental neurology
Role of the cerebrospinal fluid-contacting nucleus in the descending inhibition of spinal pain transmission.
PMID 25108066
Abstract
The brainstem is well recognized as a critical site for integrating descending modulatory systems that both inhibit and facilitate pain at the level of the spinal cord. The cerebrospinal fluid-contacting nucleus (CSF-contacting nucleus) distributes and localizes in the ventral periaqueductal central gray of the brainstem. Although emerging lines of evidence suggest that the CSF-contacting nucleus may be closely linked to transduction and regulation of pain signals, the definitive role of the CSF-contacting nucleus in pain modulation remains poorly understood. In the present study, we determined the role of the CSF-contacting nucleus in rat nocifensive behaviors after persistent pain by targeted ablation of the CSF-contacting nucleus in the brainstem using the cholera toxin subunit B-saporin (CB-SAP), a cytotoxin coupled to cholera toxin subunit B. Compared with CB/SAP, CB-SAP induced complete ablation of the CSF-contacting nucleus, and the CB-SAP-treated rats showed hypersensitivity in responses to acute nociceptive stimulation, and exacerbated spontaneous nocifensive responses induced by formalin, thermal hyperalgesia and mechanical allodynia induced by plantar incision. Furthermore, immunohistochemical experiments showed that the CSF-contacting nucleus was a cluster of 5-HT-containing neurons in the brainstem, and the spinal projection of serotonergic axons originating from the CSF-contacting nucleus constituted the descending 5-HT pathway to the spinal cord. CB-SAP induced significant downregulation of 5-HT in the spinal dorsal horn, and intrathecal injection of 5-HT significantly reversed hypersensitivity in responses to acute nociceptive stimulation in the CB-SAP-treated rats. These results indicate that the CSF-contacting nucleus 5-HT pathway is an important component of the endogenous descending inhibitory system in the control of spinal nociceptive transmission.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9523
Serotonin hydrochloride, powder
C10H12N2O · HCl