EMAIL THIS PAGE TO A FRIEND
Oncology reports
Broussonetia kazinoki modulates the expression of VEGFR-2 and MMP-2 through the inhibition of ERK, Akt and p70S6K‑dependent signaling pathways: Its implication in endothelial cell proliferation, migration and tubular formation.
PMID 25109823
Abstract
Broussonetia kazinoki (BK) has been used as a traditional medicine to improve vision, as well as for inflammatory and infectious diseases. In the present study, we investigated the effects and molecular mechanism of the ethanolic extract of BK on cell proliferation, migration and tubular formation in vascular endothelial growth factor-A (VEGF-A)-treated human umbilical vein endothelial cells. BK treatment inhibited VEGF-A-stimulated endothelial cell proliferation through the downregulation of cell cycle-related proteins including cyclin-dependent kinases and cyclins. Moreover, BK treatment suppressed cell migration and tubular formation in response to VEGF-A. These anti-angiogenic activities of BK were associated with the inactivation of mitogenic signaling pathways including extracellular signal-regulated kinase, Akt and p70S6K, and the subsequent downregulation of VEGFR-2 and matrix metalloproteinase-2. Taken together, these findings suggest further evaluation and development of BK as a potential therapeutic agent for the treatment and prevention of angiogenesis-related diseases including cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22775 Anti-AKT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501934
Anti-AKT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV36582
Anti-ANXA6 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36583
Anti-ANXA6 (AB2) antibody produced in rabbit, IgG fraction of antiserum
AV42842
Anti-ANXA6 (AB3) antibody produced in rabbit, IgG fraction of antiserum
SAB1405461
Anti-ANXA6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002462
Anti-ANXA6 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA009650
Anti-ANXA6 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1400043
Anti-CCNE1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA018169
Anti-CCNE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1400044
Anti-CCNE1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV01008 Anti-CDK2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV03006
Anti-CDK2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
C5223 Anti-Cdk2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous glycerol solution
SAB4503706
Anti-CDK2 antibody produced in rabbit, affinity isolated antibody
AV00001
Anti-CDK4 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV03017
Anti-CDK4 (AB2) antibody produced in rabbit, IgG fraction of antiserum
GW22309F Anti-CDK4 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405599
Anti-CDK4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4300695
Anti-CDK4 antibody produced in rabbit, affinity isolated antibody
SAB4503515
Anti-Cyclin E1 antibody produced in rabbit, affinity isolated antibody
SAB4503516
Anti-Cyclin E1 antibody produced in rabbit, affinity isolated antibody
AV36845
Anti-ELF1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV31085
Anti-ELF1 (AB2) antibody produced in rabbit, affinity isolated antibody
SAB2500347
Anti-ELF1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405751
Anti-ELF1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001755
Anti-ELF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100675
Anti-ELF1 antibody produced in rabbit, affinity isolated antibody
SAB1406020
Anti-IL2RB antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA062657
Anti-IL2RB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution