EMAIL THIS PAGE TO A FRIEND
Oncology reports
Prognostic significance of miR-218 in human hepatocellular carcinoma and its role in cell growth.
PMID 25110121
Abstract
MicroRNA-218 (miR-218) is considered a tumor suppressor in human cancer. In the present study, miR-218 expression was found to be significantly lower in human hepatocellular carcinoma (HCC) than in normal tumor-adjacent tissues. miR-218 was clearly silenced or downregulated in five HCC cells (HepG2, Hep3B, SMMC-7721, Huh7 and Bel-7402) compared with normal hepatocytes (LO2). The low expression of miR-218 conferred a poor 5-year survival in HCC patients. Multivariate Cox regression analysis indicated that miR-218 was an independent prognostic factor in HCC. Ectopic expression of miR-218 inhibited proliferation and promoted apoptosis in HepG2 and SMMC-7721 cells. In tumor bearing mice, miR-218 slowed down tumor growth by inducing apoptosis and growth arrest. Restoring miR-218 expression resulted in downregulation of B lymphoma Mo-MLV insertion regionxa01 homolog (BMI-1) mRNA and protein level in HepG2 and SMMC-7721 cells. In addition, BMI-1 mRNA expression in HCC was significantly higher than that in non-cancerous tissues. BMI-1 mRNA was inversely correlated with miR-218 expression in HCC tissues. In conclusion, miR-218 may serve as a prognostic biomarker and induce apoptosis and growth arrest by downregulating BMI-1 in HCC.