EMAIL THIS PAGE TO A FRIEND
Journal of clinical lipidology
Fenofibrate causes elevation of betaine excretion but not excretion of other osmolytes by healthy adults.
PMID 25110225
Abstract
Cross-sectional data suggest that bezafibrate increases betaine excretion in dyslipidemic patients. We aimed to demonstrate that fenofibrate induces increased betaine excretion in normal subjects and explore whether other 1-carbon metabolites and osmolytes are similarly affected. Urine was collected from 26 healthy adults before and after treatment with fenofibrate (145 mg/day for 6 weeks). Excretions of betaine, N,N-dimethylglycine, free choline, myo-inositol, taurine, trimethylamine-N-oxide, carnitine, and acetylcarnitine were measured by liquid chromatography with mass spectrometric detection. Fenofibrate increased the median betaine excretion from 7.5 to 25.8 mmol/mole creatinine (median increase 3-fold), P < .001. The median increase in N,N-dimethylglycine excretion was 2-fold (P < .001). Median choline excretion increased 12% (significant, P = .029). Participants with higher initial excretions tended to have larger increases (P < .001 in all 3 cases). Fenofibrate did not significantly change the median excretions of myo-inositol, taurine, trimethylamine-N-oxide, and carnitine. The excretion of acetylcarnitine decreased 4-fold on treatment, with no correlation between the baseline and after-treatment excretions. Changes in all urine components tested, except trimethylamine-N-oxide, positively correlated with changes in betaine excretion even when the median excretions before and after were not significantly different. Fibrates increase betaine, and to a lesser extent N,N-dimethylglycine and choline, excretion. Other osmolytes are not elevated. Because the increase in betaine excretion depends on the baseline excretion, large increases in excretion in the metabolic syndrome and diabetes (where baseline excretions are high) could be expected. Replacement with betaine supplements may be considered.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9500
(±)-Carnitine hydrochloride, ≥98% (TLC)
C7H16ClNO3
22020
(±)-Carnitine hydrochloride, purum, ≥99.0% (AT)
C7H16ClNO3
B3501
Betaine hydrochloride, ≥99%
C5H12ClNO2
439800
Cadmium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
CdCl2
202908
Cadmium chloride, 99.99% trace metals basis
CdCl2
655198
Cadmium chloride, technical grade
CdCl2
I5125
myo-Inositol, ≥99%
C6H12O6
I7508
myo-Inositol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
C6H12O6
57570
myo-Inositol, for microbiology, ≥99.0%
C6H12O6
RES7034I-A7
myo-Inositol, PharmaGrade, meets FCC testing specifications, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C6H12O6
Y0000485
myo-Inositol, European Pharmacopoeia (EP) Reference Standard
C6H12O6
T0625
Taurine, ≥99%
C2H7NO3S
T8691
Taurine, meets USP testing specifications, cell culture tested
C2H7NO3S
T4571
Taurine, PharmaGrade, Ajinomoto, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C2H7NO3S
PHR1109
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
C2H7NO3S
86329
Taurine, BioUltra, ≥99.5% (T)
C2H7NO3S
W381306
Taurine, ≥98%, FG
C2H7NO3S
1643361
Taurine, United States Pharmacopeia (USP) Reference Standard
C2H7NO3S
317594
Trimethylamine N-oxide, 98%
C3H9NO