EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Is rectal indomethacin effective in preventing of post-endoscopic retrograde cholangiopancreatography pancreatitis?
PMID 25110443
Abstract
To investigate the effectiveness of rectally administered indomethacin in the prophylaxis of post-endoscopic retrograde cholangiopancreatography (ERCP) pancreatitis and hyperamylasaemia in a multicentre study. A prospective, randomised, placebo-controlled multicentre study in five endoscopic units was conducted on 686 patients randomised to receive a suppository containing 100 mg indomethacin, or an inert placebo, 10-15 min before ERCP. Post-ERCP pancreatitis and hyperamylasaemia were evaluated 24 h following the procedure on the basis of clinical signs and laboratory parameters, and computed tomography/magnetic resonance imaging findings if required. Twenty-one patients were excluded because of incompleteness of their data or because of protocol violation. The results of 665 investigations were evaluated: 347 in the indomethacin group and 318 in the placebo group. The distributions of the risk factors in the two groups did not differ significantly. Pancreatitis developed in 42 patients (6.3%): it was mild in 34 (5.1%) and severe in eight (1.2%) cases. Hyperamylaesemia occurred in 160 patients (24.1%). There was no significant difference between the indomethacin and placebo groups in the incidence of either post-ERCP pancreatitis (5.8% vs 6.9%) or hyperamylasaemia (23.3% vs 24.8%). Similarly, subgroup analysis did not reveal any significant differences between the two groups. 100 mg rectal indomethacin administered before ERCP did not prove effective in preventing post-ERCP pancreatitis.