EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
CREB-induced inflammation is important for malignant mesothelioma growth.
PMID 25111229
Abstract
Malignant mesothelioma (MM) is an aggressive tumor with no treatment regimen. Previously we have demonstrated that cyclic AMP response element binding protein (CREB) is constitutively activated in MM tumor cells and tissues and plays an important role in MM pathogenesis. To understand the role of CREB in MM tumor growth, we generated CREB-inhibited MM cell lines and performed inxa0vitro and inxa0vivo experiments. Inxa0vitro experiments demonstrated that CREB inhibition results in significant attenuation of proliferation and drug resistance of MM cells. CREB-silenced MM cells were then injected into severe combined immunodeficiency mice, and tumor growth in s.c. and i.p. models of MM was followed. We observed significant inhibition in MM tumor growth in both s.c. and i.p. models and the presence of a chemotherapeutic drug, doxorubicin, further inhibited MM tumor growth in the i.p. model. Peritoneal lavage fluids from CREB-inhibited tumor-bearing mice showed a significantly reduced total cell number, differential cell counts, and pro-inflammatory cytokines and chemokines (IL-6, IL-8, regulated on activation normal T cell expressed and secreted, monocyte chemotactic protein-1, and vascular endothelial growth factor). Inxa0vitro studies showed that asbestos-induced inflammasome/inflammation activation in mesothelial cells was CREB dependent, further supporting the role of CREB in inflammation-induced MM pathogenesis. In conclusion, our data demonstrate the involvement of CREB in the regulation of MM pathogenesis by regulation of inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500441
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500442
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500443
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500444
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500445
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4300519
Anti-CREB1 (Ab-129) antibody produced in rabbit, affinity isolated antibody
SAB4300333
Anti-CREB1 (Ab-133) antibody produced in rabbit, affinity isolated antibody
SAB4300546
Anti-CREB1 (Ab-142) antibody produced in rabbit, affinity isolated antibody
AV37464
Anti-CREB1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV01026
Anti-CREB1 (AB2) antibody produced in rabbit, affinity isolated antibody
AV31264
Anti-CREB1 antibody produced in rabbit, IgG fraction of antiserum
HPA019150
Anti-CREB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103540
Anti-CREB1 antibody produced in rabbit, affinity isolated antibody
SAB4503908
Anti-phospho-CREB (pSer121) antibody produced in rabbit, affinity isolated antibody
C9102 Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504375
Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB4503757
Anti-phospho-CREB (pSer142) antibody produced in rabbit, affinity isolated antibody
SAB4504577
Anti-phospho-CREB (pThr100) antibody produced in rabbit, affinity isolated antibody
SAB1306301 ANTI-PHOSPHO-CREB(S133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300217
Anti-phospho-CREB1 (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4300040
Anti-phospho-CREB1 (pSer133) antibody produced in rabbit, affinity isolated antibody
C8977 Monoclonal Anti-CREB antibody produced in rabbit, clone 48H2, purified immunoglobulin, buffered aqueous glycerol solution
PLA0205
Rabbit anti-CREB Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.