PloS one
Generation of rat-induced pluripotent stem cells from a new model of metabolic syndrome.
PMID 25111735
Abstract
We recently characterized DahlS.Z-Leprfa/Leprfa (DS/obese) rats, derived from a cross between Dahl salt-sensitive rats and Zucker rats, as a new animal model of metabolic syndrome (MetS). Although the phenotype of DS/obese rats is similar to that of humans with MetS, the pathophysiological and metabolic characteristics in each cell type remain to be clarified. Hence, the establishment of induced pluripotent stem cells (iPSCs) derived from MetS rats is essential for investigations of MetS in vitro. Reports of rat iPSCs (riPSCs), however, are few because of the difficulty of comparing to other rodents such as mouse. Recently, the advantage of using mesenchymal stromal cells (MSCs) as a cell source for generating iPSCs was described. We aimed to establish riPSCs from MSCs in adipose tissues of both DS/obese rats and their lean littermates, DahlS.Z-Lepr+/Lepr+ (DS/lean) rats using lentivirus vectors with only three factors Oct4, Klf4, and Sox2 without c-Myc. The morphology, gene expression profiles, and protein expression of established colonies showed embryonic stem cell (ESCs)-like properties, and the differentiation potential into cells from all three germ layers both in vitro and in vivo (teratomas). Both riPSCs became adipocytes after induction of adipogenesis by insulin, T3, and dexamethasone. Real-time PCR analysis also revealed that both riPSCs and the adipose tissue from DS/obese and DS/lean rats possess similar expression patterns of adipocyte differentiation-related genes. We succeeded in generating riPSCs effectively from MSCs of both DS/obese and DS/lean rats. These riPSCs may well serve as highly effective tools for the investigation of MetS pathophysiology in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22756 Anti-Endoglin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
E7534
Anti-Endoglin antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
AV33068
Anti-ENG (AB1) antibody produced in rabbit, IgG fraction of antiserum
HPA011862
Anti-ENG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
L5037 Leptin from rat, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder
SAB4700258 Monoclonal Anti-CD105 antibody produced in mouse, clone MEM-226, purified immunoglobulin, buffered aqueous solution
SAB4700262 Monoclonal Anti-CD105 antibody produced in mouse, clone MEM-229, purified immunoglobulin, buffered aqueous solution
WH0002022M1
Monoclonal Anti-ENG antibody produced in mouse, clone 4C11, purified immunoglobulin, buffered aqueous solution
AMAB90925
Monoclonal Anti-ENG antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1912, purified immunoglobulin, buffered aqueous glycerol solution
P7280
Poly-D-lysine hydrobromide, average mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
SRP6004 Sonic Hedgehog from mouse, recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)