EMAIL THIS PAGE TO A FRIEND
Cancer letters
Gain-of-function of mutant p53: mutant p53 enhances cancer progression by inhibiting KLF17 expression in invasive breast carcinoma cells.
PMID 25111898
Abstract
Kruppel-like-factor 17 (KLF17) is a negative regulator of metastasis and epithelial-mesenchymal-transition (EMT). However, its expression is downregulated in metastatic breast cancer that contains p53 mutations. Here, we show that mutant-p53 plays a key role to suppress KLF17 and thereby enhances cancer progression, which defines novel gain-of-function (GOF) of mutant-p53. Mutant-p53 interacts with KLF17 and antagonizes KLF17 mediated EMT genes transcription. Depletion of KLF17 promotes cell viability, decreases apoptosis and induces drug resistance in metastatic breast cancer cells. KLF17 suppresses cell migration and invasion by decreasing CD44, PAI-1 and Cyclin-D1 expressions. Taken together, our results show that KLF17 is important for the suppression of metastasis and could be a potential therapeutic target during chemotherapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU123221 MISSION® esiRNA, esiRNA human TP53 (esiRNA1)