EMAIL THIS PAGE TO A FRIEND
Virology
Palmitoylation of the Alphacoronavirus TGEV spike protein S is essential for incorporation into virus-like particles but dispensable for S-M interaction.
PMID 25113909
Abstract
The spike protein S of coronaviruses contains a highly conserved cytoplasmic cysteine-rich motif adjacent to the transmembrane region. This motif is palmitoylated in the Betacoronaviruses MHV and SARS-CoV. Here, we demonstrate by metabolic labeling with [(3)H]-palmitic acid that the S protein of transmissible gastroenteritis coronavirus (TGEV), an Alphacoronavirus, is palmitoylated as well. This is relevant for TGEV replication as virus growth was compromised by the general palmitoylation inhibitor 2-bromopalmitate. Mutation of individual cysteine clusters in the cysteine-rich motif of S revealed that all cysteines must be replaced to abolish acylation and incorporation of S into virus-like particles (VLP). Conversely, the interaction of S with the M protein, essential for VLP incorporation of S, was not impaired by lack of palmitoylation. Thus, palmitoylation of the S protein of Alphacoronaviruses is dispensable for S-M interaction, but required for the generation of progeny virions.