EMAIL THIS PAGE TO A FRIEND
Oncotarget
Aurora kinase a suppresses metabolic stress-induced autophagic cell death by activating mTOR signaling in breast cancer cells.
PMID 25115395
Abstract
Aberrant Aur-A signaling is associated with tumor malignant behaviors. However, its involvement in tumor metabolic stress is not fully elucidated. In the present study, prolonged nutrient deprivation was conducted into breast cancer cells to mimic metabolic stress in tumors. In these cells, autophagy was induced, leading to caspase-independent cell death, which was blocked by either targeted knockdown of autophagic gene ATG5 or autophagy inhibitor 3-Methyladenine (3-MA). Aur-A overexpression mediated resistance to autophagic cell death and promoted breast cancer cells survival when exposed to metabolic stress. Moreover, we provided evidence that Aur-A suppressed autophagy in a kinase-dependent manner. Furthermore, we revealed that Aur-A overexpression enhanced the mammalian target of rapamycin (mTOR) activity under metabolic stress by inhibiting glycogen synthase kinase 3β (GSK3β). Inhibition of mTOR activity by rapamycin sensitized Aur-A-overexpressed breast cancer cells to metabolic stress-induced cell death. Consistently, we presented an inverse correlation between Aur-A expression (high) and autophagic levels (low) in clinical breast cancer samples. In conclusion, our data provided a novel insight into the cyto-protective role of Aur-A against metabolic stress by suppressing autophagic cell death, which might help to develop alternative cell death avenues for breast cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-648
Anti-Aurora A Antibody, 200 µg, Detect Aurora A with Anti-Aurora A Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB.
06-734
Anti-β-Catenin Antibody, 200 µg, Use Anti-β-Catenin Antibody (Rabbit Polyclonal Antibody) validated in ICC, IP, WB to detect β-Catenin also known as catenin (cadherin-associated protein) beta 1 (88kD).
02685
Lithium atomic spectroscopy standard concentrate 1.00 g Li, for 1 l standard solution, 1.00 g/L, analytical standard
ClLi
746460
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
ClLi
793620
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
ClLi
L4408
Lithium chloride, BioXtra, ≥99.0% (titration)
ClLi
62476
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
ClLi
L9650
Lithium chloride, for molecular biology, ≥99%
ClLi
429457
Lithium chloride, anhydrous, beads, −10 mesh, 99.998% trace metals basis
ClLi
449881
Lithium chloride, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
ClLi
203637
Lithium chloride, powder, ≥99.99% trace metals basis
ClLi
HT-LICLSLUG
Lithium chloride, anhydrous, chunks, 99.99% trace metals basis
ClLi
751898
Lithium chloride, anhydrous, chunks, 99.99% trace metals basis
ClLi
L7026
Lithium chloride solution, 8 M, for molecular biology, ≥99%
ClLi
694576
Lithium chloride solution, 1 M in 1,3-dimethyl-2-imidazolidinone
ClLi
62483
Lithium chloride solution, 1 M in glacial acetic acid
ClLi
62481
Lithium chloride solution, 1 M in ethanol
ClLi
667544
Lithium chloride solution, 0.5 M in anhydrous tetrahydrofuran
ClLi
LCL30
Lithium chloride solution, 2 M in ethanol
ClLi
601489
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Cl7Li
37094
Rapamycin, VETRANAL, analytical standard
C51H79NO13