EMAIL THIS PAGE TO A FRIEND
Virus research
Sequences outside that of residues 93-102 of 3A protein can contribute to the ability of foot-and-mouth disease virus (FMDV) to replicate in bovine-derived cells.
PMID 25116389
Abstract
Foot-and-mouth disease (FMD) is a highly contagious and economically devastating disease of cloven-hoofed animals. During 2010 and 2011, there was an epidemic of the Mya-98 lineage of the Southeast Asia (SEA) topotype in East Asia, including China. Changes in the FMDV 3A protein have been previously reported to be associated with the inability of FMDV to grow in bovine cells and cause disease in cattle. In this paper, we report the generation of a full-length infectious cDNA clone of FMDV O/SEA/Mya-98 strain O/GZSB/2011 for the first time along with two genetically modified viruses with deletion at positions 93-102 and 133-143 in 3A based on the established infectious clone. All the recombinant viruses grew well and displayed growth properties and plaque phenotypes similar to those of the parental virus in baby hamster kidney (BHK-21) cells, porcine kidney (PK-15) cells, and primary fetal porcine kidney (FPK) cells. While the recombinant viruses rvGZSB and rvSBΔ133-143 exhibited similar growth properties and plaque phenotypes with the parental virus in primary fetal bovine kidney (FBK) cells, the recombinant virus rvSBΔ93-102, containing deletion at positions 93-102 in 3A, grew at a slower rate and had a smaller plaque size phenotype in FBK cells than that of the parental virus. Therefore, the results suggest that the deletion at positions 93-102 of 3A protein does not affect FMDV replication efficiency in BHK-21, PK-15 and FPK cells, but affects virus replication efficiency in FBK cells, although, cannot alone account for the inability to replicate in bovine cells.