EMAIL THIS PAGE TO A FRIEND
Cancer immunology research
A feasibility study of cyclophosphamide, trastuzumab, and an allogeneic GM-CSF-secreting breast tumor vaccine for HER2+ metastatic breast cancer.
PMID 25116755
Abstract
Granulocyte-macrophage colony-stimulating factor (GM-CSF)-secreting tumor vaccines are bioactive, but limited by disease burden and immune tolerance. Cyclophosphamide augments vaccine activity in tolerant neu mice and in patients with metastatic breast cancer. HER2-specific monoclonal antibodies (mAb) enhance vaccine activity in neu mice. We hypothesized that cyclophosphamide-modulated vaccination with HER2-specific mAb safely induces relevant HER2-specific immunity in neu mice and patients with HER2+ metastatic breast cancer. Adding both cyclophosphamide and the HER2-specific mAb 7.16.4 to vaccination maximized HER2-specific CD8+ T-cell immunity and tumor-free survival in neu transgenic mice. We, therefore, conducted a single-arm feasibility study of cyclophosphamide, an allogeneic HER2+ GM-CSF-secreting breast tumor vaccine, and weekly trastuzumab in 20 patients with HER2+ metastatic breast cancer. Primary clinical trial objectives were safety and clinical benefit, in which clinical benefit represents complete response + partial response + stable disease. Secondary study objectives were to assess HER2-specific T-cell responses by delayed type hypersensitivity (DTH) and intracellular cytokine staining. Patients received three monthly vaccinations, with a boost 6 to 8 months from trial entry. This combination immunotherapy was safe, with clinical benefit rates at 6 months and 1 year of 55% [95% confidence interval (CI), 32%-77%; P = 0.013] and 40% (95% CI, 19%-64%), respectively. Median progression-free survival and overall survival durations were 7 months (95% CI, 4-16) and 42 months (95% CI, 22-70), respectively. Increased HER2-specific DTH developed in 7 of 20 patients [of whom 4 had clinical benefit (95% CI, 18-90)], with a trend toward longer progression-free survival and overall survival in DTH responders. Polyfunctional HER2-specific CD8+ T cells progressively expanded across vaccination cycles. Further investigation of cyclophosphamide-modulated vaccination with trastuzumab is warranted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3250000
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard
C7H15Cl2N2O2P · H2O