EMAIL THIS PAGE TO A FRIEND
Biotechnology and bioengineering
Scaled-up manufacturing of recombinant antibodies produced by plant cells in a 200-L orbitally-shaken disposable bioreactor.
PMID 25117428
Abstract
Tobacco BY-2 cells have emerged as a promising platform for the manufacture of biopharmaceutical proteins, offering efficient protein secretion, favourable growth characteristics and cultivation in containment under a controlled environment. The cultivation of BY-2 cells in disposable bioreactors is a useful alternative to conventional stainless steel stirred-tank reactors, and orbitally-shaken bioreactors could provide further advantages such as simple bag geometry, scalability and predictable process settings. We carried out a scale-up study, using a 200-L orbitally-shaken bioreactor holding disposable bags, and BY-2 cells producing the human monoclonal antibody M12. We found that cell growth and recombinant protein accumulation were comparable to standard shake flask cultivation, despite a 200-fold difference in cultivation volume. Final cell fresh weights of 300-387 g/L and M12 yields of ∼20 mg/L were achieved with both cultivation methods. Furthermore, we established an efficient downstream process for the recovery of M12 from the culture broth. The viscous spent medium prevented clarification using filtration devices, but we used expanded bed adsorption (EBA) chromatography with SP Sepharose as an alternative for the efficient capture of the M12 antibody. EBA was introduced as an initial purification step prior to protein A affinity chromatography, resulting in an overall M12 recovery of 75-85% and a purity of >95%. Our results demonstrate the suitability of orbitally-shaken bioreactors for the scaled-up cultivation of plant cell suspension cultures and provide a strategy for the efficient purification of antibodies from the BY-2 culture medium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1000000
Methotrexate, European Pharmacopoeia (EP) Reference Standard
C20H22N8O5
M7824
Methotrexate, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, meets EP, USP testing specifications
C20H22N8O5
PHR1396
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
C20H22N8O5
M4010
Methotrexate, meets EP, USP testing specifications
C20H22N8O5 · xH2O
M9929
Methotrexate, meets USP testing specifications
C20H22N8O5 · xH2O
Y0000602
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
C20H22N8O5
Y0001668
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
C20H22N8O5
791741
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C2H3NaO2
S2889
Sodium acetate, anhydrous, for molecular biology, ≥99%
C2H3NaO2
S3272
Sodium acetate, for electrophoresis, ≥99%, crystalline
C2H3NaO2
S5636
Sodium acetate, powder, BioReagent, for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
C2H3NaO2
S1429
Sodium acetate, meets USP testing specifications, anhydrous
C2H3NaO2
1613407
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
C2H3NaO2
229873
Sodium acetate, 99.995% trace metals basis
C2H3NaO2
S7545
Sodium acetate, BioXtra, ≥99.0%
C2H3NaO2
110191
Sodium acetate, ReagentPlus®, 99%
C2H3NaO2
71183
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
C2H3NaO2
W302406
Sodium Acetate Anhydrous, >99%, FCC, FG
C2H3NaO2
S8388
Sodium acetate solution, 0.3 M
C2H3NaO2
71196
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
C2H3NaO2
613053
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
C2H3NaO2