EMAIL THIS PAGE TO A FRIEND
European radiology
Volumetric MR-guided high-intensity focused ultrasound versus uterine artery embolisation for treatment of symptomatic uterine fibroids: comparison of symptom improvement and reintervention rates.
PMID 25117743
Abstract
To compare the effectiveness of magnetic resonance-guided high-intensity focused ultrasound (MR-HIFU) with that of uterine artery embolisation (UAE) for treatment of uterine fibroids. Between January 2010 and January 2013, 51 women with symptomatic uterinexa0fibroids underwent MR-HIFU. Follow-up and MR imaging were compared to 68 women treated with UAE, who fulfilled eligibility criteria for MR-HIFU - e.g., size (≤xa012xa0cm) and number (≤xa05)xa0of fibroids. We compared median symptom severity (tSSS), totalxa0health-realted qualityxa0ofxa0life (HRQoL) scores, and reintervention rates. The adjusted effect on symptom relief and HRQoL improvement was calculated using multivariable linear regression. Cox regression was applied to calculate the adjusted risk of reintervention between both treatments. Median tSSS improved significantly from baseline to three-month follow-up (P < 0.001) for both MR-HIFU (53.1 (IQR [40.6-68.8]) to 34.4 (IQR [21.9-46.9]) and UAE (65.3 (IQR [56.3-74.2]) to 21.9 (IQR [9.4-34.4]). In addition, significantly better HRQoL scores were observed after three months (P < 0.001). However, in multivariate analysis, UAE had a stronger effect on symptom relief and HRQoLxa0improvement than MR-HIFU (P < 0.001). Patients treated with MR-HIFU had a 7.1 (95xa0% CI [2.00-25.3]; P = 0.002) times higher risk of reintervention within 12xa0months (18/51 vs. 3/68). Both MR-HIFU and UAE result in significant symptom relief related to uterine fibroids. However, MR-HIFU is associated with a higher risk of reintervention. • This study compared outcomes between volumetric MR-HIFU and UAE for uterinexa0fibroids. • Both MR-HIFU and UAE result in significant symptom relief and qualityxa0ofxa0life improvement. • UAE had a stronger positive effect on thexa0clinical outcomes. • Reintervention rate after MR-HIFU ablation was significantly higher than after UAE.