EMAIL THIS PAGE TO A FRIEND
Reproduction (Cambridge, England)
CAR expression in human embryos and hESC illustrates its role in pluripotency and tight junctions.
PMID 25118298
Abstract
Coxsackie virus and adenovirus receptor, CXADR (CAR), is present during embryogenesis and is involved in tissue regeneration, cancer and intercellular adhesion. We investigated the expression of CAR in human preimplantation embryos and embryonic stem cells (hESC) to identify its role in early embryogenesis and differentiation. CAR protein was ubiquitously present during preimplantation development. It was localised in the nucleus of uncommitted cells, from the cleavage stage up to the precursor epiblast, and corresponded with the presence of soluble CXADR3/7 splice variant. CAR was displayed on the membrane, involving in the formation of tight junction at compaction and blastocyst stages in both outer and inner cells, and CAR corresponded with the full-length CAR-containing transmembrane domain. In trophectodermal cells of hatched blastocysts, CAR was reduced in the membrane and concentrated in the nucleus, which correlated with the switch in RNA expression to the CXADR4/7 and CXADR2/7 splice variants. The cells in the outer layer of hESC colonies contained CAR on the membrane and all the cells of the colony had CAR in the nucleus, corresponding with the transmembrane CXADR and CXADR4/7. Upon differentiation of hESC into cells representing the three germ layers and trophoblast lineage, the expression of CXADR was downregulated. We concluded that CXADR is differentially expressed during human preimplantation development. We described various CAR expressions: i) soluble CXADR marking undifferentiated blastomeres; ii) transmembrane CAR related with epithelial-like cell types, such as the trophectoderm (TE) and the outer layer of hESC colonies; and iii) soluble CAR present in TE nuclei after hatching. The functions of these distinct forms remain to be elucidated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4362
Activin A active human, Animal-component free, recombinant, expressed in Nicotiana, >97% (SDS-PAGE)
A4941 Activin A human, >95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, lyophilized powder, cell culture tested
H4666
Activin A human, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
SRP6153
Activin A human, recombinant, expressed in human cells, ≥95% (SDS-PAGE)
SRP3003 Activin A human, recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC), cell culture tested
HPA030411
Anti-CXADR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
SAB1406198
Anti-OCLN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101697
Anti-OTX2 (ab2) antibody produced in rabbit, affinity isolated antibody
HPA000633
Anti-OTX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32439
Anti-OTX2 antibody produced in rabbit, affinity isolated antibody
AV48225
Anti-VIM (AB1) antibody produced in rabbit, affinity isolated antibody
AV48226
Anti-VIM (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1406585
Anti-VIM antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001762
Anti-VIM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
V4630 Anti-Vimentin antibody produced in goat, whole antiserum
SAB4300676
Anti-Vimentin antibody produced in rabbit, affinity isolated antibody
SAB4503081
Anti-Vimentin antibody produced in rabbit, affinity isolated antibody
SRP3260
BMP-4 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
SRP6156
BMP-4 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
SRP3016
BMP-4 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
WH0004950M1
Monoclonal Anti-OCLN antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution
AMAB90893
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1608, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90890
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1567, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90889
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1555, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300043
Monoclonal Anti-OTX2 antibody produced in mouse, clone 1H12C4B5, ascites fluid
SAB5300044
Monoclonal Anti-OTX2 antibody produced in mouse, clone 1H12G8B2, ascites fluid
WH0007431M1
Monoclonal Anti-VIM antibody produced in mouse, clone 3E9, purified immunoglobulin, buffered aqueous solution
SAB4700524 Monoclonal Anti-VIM antibody produced in mouse, clone VI-RE/1, purified immunoglobulin, buffered aqueous solution
SAB4100612
Monoclonal Anti-VIM antibody produced in mouse, clone VIM-A604, culture supernatant
AMAB90516
Monoclonal Anti-VIM antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0157, purified immunoglobulin, buffered aqueous glycerol solution