EMAIL THIS PAGE TO A FRIEND
Life sciences
The effects of PTBP3 silencing on the proliferation and differentiation of MKN45 human gastric cancer cells.
PMID 25119103
Abstract
PTBP3 overexpression inhibits the differentiation of leukemia cells; however, its effects on the differentiation and proliferation of solid cancer cells remain unclear. Thus, the impact of PTBP3 on the differentiation and proliferation of gastric cancer cells was investigated. PTBP3 expression was analyzed in normal and tumor tissues using immunohistochemistry. A xenograft model was established in nude mice by subcutaneous injection of untransfected human gastric cancer MKN45 cells or those expressing a control vector or PTBP3 siRNA. We analyzed the tumor inhibition rate, the expression of PTBP3, the PCNA-positive rate and the serum levels of CEA, CA199, CA125, LDH, ALP and γ-GT in different groups. The tumor weights in the PTBP3 siRNA group were significantly lower than that of the MKN45 cell control group (P<0.001). Immunohistochemistry analysis of PCNA expression revealed that it was markedly reduced after PTBP3 silencing. ELISAs showed that the serum levels of CEA and CA199 tumor markers as well as LDH and ALP were reduced after PTBP3 silencing. Transmission electron microscopy revealed that MKN45 cells expressing PTBP3 siRNA had reduced nuclear-to-cytoplasmic ratio and regular nuclei, suggesting differentiation. PTBP3 may promote proliferation and inhibit the differentiation of human gastric cancer MKN45 cells.