EMAIL THIS PAGE TO A FRIEND
Nanotechnology
Immunoadjuvant activity of the nanoparticles' surface modified with mannan.
PMID 25119543
Abstract
Mannan (MN) is the natural ligand for mannose receptors, which are widely expressed on dendritic cells (DCs). The purpose of this study was to assess the effect of formulation parameters on the immunogenicity of MN-decorated poly (D, L-lactide-co-glycolide) (PLGA) nanoparticles (NPs) in terms of their ability to stimulate DC phenotypic as well as functional maturation. For this purpose, NPs were formulated from either ester-terminated or COOH-terminated PLGA. Incorporation of MN in NPs was achieved through encapsulation, physical adsorption or chemical conjugation. Murine bone marrow derived DCs (BMDCs) were treated with various NP formulations and assessed for their ability to up-regulate DC cell surface markers, secrete immunostimulatory cytokines and to activate allogenic T cell responses. DCs treated with COOH-terminated PLGA-NPs containing chemically conjugated MN (MN-Cov-COOH) have shown superior performance in improving DC biological functions, compared to the rest of the formulations tested. This may be attributed to the higher level of MN incorporation in the former formulation. Incorporation of MN in PLGA NPs through chemical conjugation can lead to enhanced DC maturation and stimulatory function. This strategy may be used to develop more effective PLGA-based vaccine formulations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV41929
Anti-CD40 antibody produced in rabbit, affinity isolated antibody
HPA031567
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031568
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103449
Anti-CD40, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1411654
Anti-CD86 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
RAB0521
Bovine IL2 / Interleukin-2 ELISA Kit, for serum, plasma and cell culture supernatants
RAB0286
Human IL-2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
WH0000958M1
Monoclonal Anti-CD40 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
AMAB90905
Monoclonal Anti-CD40 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1673, purified immunoglobulin, buffered aqueous glycerol solution
WH0000942M1 Monoclonal Anti-CD86 antibody produced in mouse, clone 3D7, purified immunoglobulin, buffered aqueous solution
SAB4700144 Monoclonal Anti-CD86 antibody produced in mouse, clone BU63, purified immunoglobulin, buffered aqueous solution
SAB4700560 Monoclonal Anti-Cd86 antibody produced in rat, clone GL-1, purified immunoglobulin, buffered aqueous solution
RAB0287
Mouse IL-2 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0288
Rat IL-2 ELISA Kit, for serum, plasma, cell culture supernatant