EMAIL THIS PAGE TO A FRIEND
Journal of human hypertension
Large artery stiffness and carotid intima-media thickness in relation to markers of calcium and bone mineral metabolism in African women older than 46 years.
PMID 25119885
Abstract
Vascular calcification and cardiovascular diseases have been associated with altered bone metabolism. We explored the relationships of arterial pressures and carotid intima-media thickness (CIMT) with parathyroid hormone, 25-hydroxycholecalciferol and their ratio (PTH:25(OH)D3) as well as a marker of bone resorption (CTX) in lean and overweight/obese African women. A population of 434 African women older than 46 years was divided into lean and overweight/obese groups. We assessed brachial blood pressure, central pulse pressure (cPP) and CIMT, and determined PTH, 25(OH)D3 and CTX concentrations. Overweight/obese women had elevated PTH and PTH:25(OH)D3 compared with lean women (both P<0.001), whereas lean women had higher CTX (P<0.001). Single, partial and multiple regression analyses indicated that, in lean women CIMT was independently associated with PTH:25(OH)D3 (R(2)=0.22; β=0.26; P=0.003), whereas in obese women cPP was associated with both PTH:25(OH)D3 (R2=0.20; β=0.17; P=0.017) and CTX (R2=0.20; β=0.17; P=0.025). In conclusion, we found that in African women with increased adiposity, cPP (as a surrogate measure of arterial stiffness), was positively associated with alterations in bone metabolism and calciotropic hormones, whereas CIMT of lean women was positively associated with PTH:25(OH)D3. Our results suggest that alterations in bone and calcium metabolism may contribute to arterial calcification in older African women.