EMAIL THIS PAGE TO A FRIEND
European radiology
Correlations of (18)F-fluorothymidine uptake with pathological tumour size, Ki-67 and thymidine kinase 1 expressions in primary and metastatic lymph node colorectal cancer foci.
PMID 25120206
Abstract
To examine correlations of (18)F-fluorothymidine (FLT) uptake with pathological tumour size and immunohistochemical Ki-67, and thymidine kinase 1 (TK-1) expressions in primary and metastatic node colorectal cancer foci. Thirty primary cancers (PCs) and 37 metastatic nodes (MNs) were included. FLT uptake was assessed by visual scores (non-visible: 0-1 and visible: 2-4), standardized uptake value (SUV), and correlated with size, Ki-67, and TK-1. SUV was measured in visible lesions. FLT heterogeneity was assessed by visual scores (no heterogeneous uptake: 0 and heterogeneous uptake: 1-4). Forty-two lesions were visible. The visible group showed significantly higher values than the non-visible group in size, Ki-67, and TK-1 (each p < 0.05). Size correlated significantly with visual score (PC; ρ = 0.74 and MN; ρ = 0.63), SUVmax (PC; ρ = 0.49, and MN; ρ = 0.76), and SUVmean (PC; ρ = 0.40 and MN; ρ = 0.76) (each p < 0.05). Visual score correlated significantly with size (ρ = 0.86), Ki-67max (ρ = 0.35), Ki-67mean (ρ = 0.38), TK-1max (ρ = 0.35) and TK-1mean (ρ = 0.25) (each p < 0.05). No significant correlations were found between FLT uptake and Ki-67 or TK-1 in 42 visible lesions (each p > 0.05). Heterogeneous FLT uptake was noted in 73 % (22/30) of PCs. FLT uptake correlated with size. Heterogeneous FLT distribution in colorectal cancers may be one of the causes of weak or lack of FLT uptake/Ki-67 or TK-1 correlation. FLT uptake correlated well with tumour size in colorectal cancer. Weak or lack of FLT uptake/Ki-67 and TK-1 correlations were observed. Immunohistochemical Ki-67 and TK-1 expressions are not always correlated with FLT uptake.