EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Disuse deterioration of human skeletal muscle challenged by resistive exercise superimposed with vibration: evidence from structural and proteomic analysis.
PMID 25122557
Abstract
In the present bed rest (BR) study, 23 volunteers were randomized into 3 subgroups: 60 d BR control (Ctr); BR with resistive exercise (RE; lower-limb load); and resistive vibration exercise (RVE; RE with superimposed vibration). The aim was to analyze by confocal and electron microscopy the effects of vibration on myofibril and filament integrity in soleus (Sol) and vastus lateralis (VL) muscle; differential proteomics of contractile, cytoskeletal, and costameric proteins (TN-C, ROCK1, and FAK); and expression of PGC1α and atrophy-related master genes MuRF1 and MuRF2. RVE (but not RE) preserved myofiber size and phenotype in Sol and VL by overexpressing MYBPC1 (42%, P ≤ 0.01), WDR1 (39%, P ≤ 0.01), sarcosin (84%, P ≤ 0.01), and CKM (20%, P ≤ 0.01) and prevented myofibrillar ultrastructural damage as detectable by MuRF1 expression. In Sol, cytoskeletal and contractile proteins were normalized by RVE, and TN-C increased (59%, P ≤ 0.01); the latter also with RE (108%, P ≤ 0.01). In VL, the outcomes of both RVE (acting on sarcosin and desmin) and RE (by way of troponinT-slow and MYL2) were similar. RVE appears to be a highly efficient countermeasure protocol against muscle atrophy and ultrastructural and molecular dysregulation induced by chronic disuse.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09972
3-Nitrotetrazolium blue chloride, ≥75% (AT)
C40H30Cl2N10O6
HPA007567
Anti-ROCK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA045639
Anti-ROCK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21017 Anti-Tenascin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA004823
Anti-TNC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1409900
Anti-TRIM63 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2105510
Anti-TRIM63 antibody produced in rabbit, affinity isolated antibody
SAB1411517
Anti-TRIM63 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0006093M1
Monoclonal Anti-ROCK1 antibody produced in mouse, clone 2E2, purified immunoglobulin, buffered aqueous solution
SAB1404324
Monoclonal Anti-ROCK1 antibody produced in mouse, clone 2E2, ascites fluid
SAB5300386
Monoclonal Anti-ROCK1 antibody produced in mouse, clone 1H4, purified immunoglobulin, buffered aqueous solution
T2551 Monoclonal Anti-Tenascin antibody produced in mouse, clone BC-24, ascites fluid
T3413 Monoclonal Anti-Tenascin antibody produced in rat, clone MTn-12, ascites fluid
WH0003371M1
Monoclonal Anti-TNC antibody produced in mouse, clone 3B4, purified immunoglobulin, buffered aqueous solution
WH0084676M1
Monoclonal Anti-TRIM63 antibody produced in mouse, clone 6G6, purified immunoglobulin, buffered aqueous solution
T4375
Tetrazolium Blue Chloride, used in colorimetric determination of reducing compounds
C40H32Cl2N8O2
88190
Tetrazolium Blue Chloride, indicator for germination, for microbiology, ≥90% (T)
C40H32Cl2N8O2