EMAIL THIS PAGE TO A FRIEND
Journal of virology
High frequencies of polyfunctional CD8+ NK cells in chronic HIV-1 infection are associated with slower disease progression.
PMID 25122796
Abstract
Natural killer (NK) cells are effector and regulatory innate immune cells and play a critical role in the first line of defense against various viral infections. Although previous reports have indicated the vital contributions of NK cells to HIV-1 immune control, nongenetic NK cell parameters directly associated with slower disease progression have not been defined yet. In a longitudinal, retrospective study of 117 untreated HIV-infected subjects, we show that higher frequencies as well as the absolute numbers of CD8(+) CD3(-) lymphocytes are linked to delayed HIV-1 disease progression. We show that the majority of these cells are well-described blood NK cells. In a subsequent cross-sectional study, we demonstrate a significant loss of CD8(+) NK cells in untreated HIV-infected individuals, which correlated with HIV loads and inversely correlated with CD4(+) T cell counts. CD8(+) NK cells had modestly higher frequencies of CD57-expressing cells than CD8(-) cells, but CD8(+) and CD8(-) NK cells showed no differences in the expression of a number of activating and inhibiting NK cell receptors. However, CD8(+) NK cells exhibited a more functional profile, as detected by cytokine production and degranulation. We demonstrate that the frequency of highly functional CD8(+) NK cells is inversely associated with HIV-related disease markers and linked with delayed disease progression. These results thus indicate that CD8(+) NK cells represent a novel NK cell-derived, innate immune correlate with an improved clinical outcome in HIV infection.